Cell Type: | HEK293 |
---|---|
Concentration: | 10 x solution |
Product Line: | AAV-MAX |
Reagent Type: | cell lysis buffer |
Shipping Condition: | wet ice |
Composition: | Polysorbate 20-based |
For Use With (Application): | AAV Production |
Sufficient For: | 1 L of culture |