Pierce™ High-Capacity EDTA-Compatible Ni-IMAC Resin
Thermo Scientific™

Pierce™ High-Capacity EDTA-Compatible Ni-IMAC Resin

Pierce High-Capacity EDTA-Compatible Ni-IMAC Resin is designed for the purification of over-expressed His-tagged proteins from cell culture media.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A5058410 mL
A5058550 mL
A50586250 mL
A50587500 mL
A400045635 Columns
A400045652 Plates
Catalog number A50584
Price (USD)
164.65
Online exclusive
185.00
Save 20.35
Each
Quantity:
10 mL
Price (USD)
164.65
Online exclusive
185.00
Save 20.35
Each

Pierce High-Capacity EDTA-Compatible Ni-IMAC Resin is designed for the purification of over-expressed His-tagged proteins from cell culture media. The matrix is a 6% crosslinked agarose with optimal properties for batch, column-based, and research/pilot scale FPLC column purifications. The resin has high binding capacity (up to 80 mg of protein per mL of resin) and can tolerate the high EDTA and reducing agent concentrations used to inhibit metalloproteases and stabilize sensitive intracellular proteins.

Features of Pierce high-Capacity EDTA-Compatible Ni-IMAC Resin

  • Chelator stable—use up to 20 mM EDTA and DTT with no loss in performance
  • Versatile—purify both overexpressed and secreted proteins in cell culture media (e.g., Expi293, ExpiCHO, and ExpiSf expression systems)
  • High binding capacity—>80 mg/mL binding capacity for His-tagged recombinant proteins
  • Reusable—loose resin and spin column formats can be regenerated up to 10 times using the standard CIP protocol (not applicable to 96-well plates)
  • Flexible— available in multiple formats including bulk loose resin, spin columns, and 96-well spin plates

EDTA and reducing agents such as DTT and TCEP adversely affect the performance of Ni-IMAC supports by stripping off the metal. Because of its chelator stability characteristic (EDTA tolerant), Pierce High-Capacity EDTA-Compatible Ni-IMAC Resin is specially suited for purification of expressed His-tagged proteins that are secreted into cell culture media or intracellular His-tagged proteins that need the presence of EDTA to maintain stability and function. In both cases, large quantities of samples can be purified without the Ni metal stripping off of the support, thereby providing consistent high-capacity yields.

Available formats

  • Resin slurries—20 mL, 100 mL, 500 mL, and 1 L of 50% slurry in 20% ethanol
  • Spin columns—10 mL spin columns containing 1 mL of settled resin, supplied as 5 mL of 20% slurry
  • 96-well spin plates—96-well spin plates with 50 μL of settled resin, supplied as 250 μL of 20% slurry
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Binding PropertyChelator Stable
Column TypeBottle
Matrix6% crosslinked agarose
Quantity10 mL
Shipping ConditionWet Ice
Binding Capacity80 mg/mL Binding
FormLiquid Suspension
Particle Size50 to 80 μm
Product LinePierce
TypeResin
Unit SizeEach
Contents & Storage
10 mL settled resin, supplied as 20 mL 50% slurry. Store at 2–8°C.

Frequently asked questions (FAQs)

With Pierce High Capacity Ni-IMAC Resin, EDTA compatible, what residence times are adequate for FPLC purification?

Optimized residence times will differ by target, but the standard range is 2-10 mins.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

How is Pierce High Capacity Ni-IMAC Resin, EDTA compatible different from standard Ni-NTA resin?

Pierce High Capacity Ni-IMAC Resin, EDTA compatible doesn't leach and discolor in the presence of reducing agent, EDTA, or cell culture supernatant, thereby providing exceptionally high protein yield.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

After regeneration of Pierce High Capacity Ni-IMAC Resin, EDTA compatible, is there a specific buffer the resin should be stored in?

The recommended storage buffer for the column and resin is 20% ethanol, 10 mM sodium acetate, pH 6.50. Please refer to the product manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0024862_PierceHighCapNiIMACResin_UG.pdf) for details.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

With Pierce High Capacity Ni-IMAC Resin, EDTA compatible, how do I regenerate the resin?

Pierce High Capacity Ni-IMAC Resin, EDTA compatible is alkali-stable. The resin can be exposed to NaOH, washed with water, neutralized with PBS, washed with water, and put back into storage buffer. Please refer to the product manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0024862_PierceHighCapNiIMACResin_UG.pdf) for details.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

How many uses/regenerations are possible with Pierce High Capacity Ni-IMAC Resin, EDTA compatible?

Up to 10 uses are possible with Pierce High Capacity Ni-IMAC Resin, EDTA compatible.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.