Spectral Calibration Plate with Alexa Fluor 647 Dye, 96-well Fast
Thermo Scientific™

Spectral Calibration Plate with Alexa Fluor 647 Dye, 96-well Fast

The 96-well Fast Spectral Calibration Plate with Alexa Fluor 647 is used to maintain the QuantStudio 5 Dx and QuantStudio 7 Pro Dx real-time PCR instruments with 96-well 0.1-mL (Fast) blocks.
Have Questions?
Catalog NumberQuantity
A505961 Plate
Catalog number A50596
Price (USD)
213.00
Each
Quantity:
1 Plate
Price (USD)
213.00
Each

The 96-well Fast Spectral Calibration Plate with Alexa Fluor 647 is used to maintain the QuantStudio 5 Dx and QuantStudio 7 Pro Dx real-time PCR instruments with 96-well 0.1-mL (Fast) blocks. Please see the instrument user guide for full instructions on use.

For In Vitro Diagnostic Use. Not available in all countries or regions.
Specifications
Calibrated DyeAlexa Fluor 647
Calibration TypePure Dye Calibration
For Use With (Application)Pure Dye Calibration
For Use With (Equipment)7500 Fast Dx Real‐Time PCR Instrument
Format96-well, 0.1 mL
Quantity1 Plate
Shelf Life12 Months
Shipping ConditionDry Ice
Product TypePlate
Unit SizeEach
Contents & Storage
Store at -20°C ±5°C.