Oncomine™ Myeloid RNA Assay GX v2
Oncomine™ Myeloid RNA Assay GX v2
Ion Torrent™

Oncomine™ Myeloid RNA Assay GX v2

The Ion Torrent Oncomine Myeloid RNA Assay GX v2 is a comprehensive targeted next-generation sequencing (NGS) assay designed for sensitiveRead more
Have Questions?
Catalog NumberQuantity
A5075432 samples kit
Catalog number A50754
Price (USD)
6,800.00
Each
Add to cart
Quantity:
32 samples kit
Price (USD)
6,800.00
Each
Add to cart
The Ion Torrent Oncomine Myeloid RNA Assay GX v2 is a comprehensive targeted next-generation sequencing (NGS) assay designed for sensitive detection of myeloid disorder-associated RNA fusion transcripts in blood and bone marrow samples. This assay is the RNA component of the Oncomine Myeloid Assay GX v2 (Cat. No. A50694), which includes both DNA and RNA assays. This assay is compatible with the Genexus Integrated Sequencer, which performs library preparation, sequencing, analysis, and reporting in an automated sample-to-result workflow. Depending on the workflow, results can be obtained in as little as a single day.

The Oncomine Myeloid RNA Assay GX v2 features include:
• Comprehensive coverage of >700 key fusion transcripts associated with myeloid disorders
• Automated sample-to-report workflow on the Genexus sequencer in less than a day
• Sequence up to 32 RNA samples on one lane of a GX5 Chip in a single run
• Less than 15 minutes hands-on time

The Oncomine Myeloid RNA Assay GX v2 is designed to assist in myeloid cancer research. The RNA panel includes 30 fusion driver genes covering the most relevant targets associated with major myeloid disorders, including acute myeloid leukemia (AML), myeloid dysplastic syndrome (MDS), myeloproliferative neoplasms (MPN), chronic myeloid leukemia (CML), chronic myelomonocytic leukemia (CMML), and juvenile myelomonocytic leukemia (JMML).

The multiplex primer design leverages Ion AmpliSeq technology to generate results from multiple samples in a single run. Sequencing results are automatically analyzed by Genexus software using an optimized assay-specific analysis workflow.

The assay kit includes one pool of Ion AmpliSeq oligonucleotide primers and Genexus Strips 1 & 2-AS library reagents, enough to sequence up to 32 RNA samples on the Genexus Integrated Sequencer. One end-to-end sequencing run uses one lane on the GX5 Chip and accommodates up to 32 samples. Additional Genexus sequencer reagents and supplies must be ordered separately.
Specifications
For Use With (Application)Sequencing
For Use With (Equipment)Genexus™ Integrated Sequencer
No. of Reactions32
Quantity32 samples kit
Sample TypeRNA from Blood or Bone Marrow
Unit SizeEach

Frequently asked questions (FAQs)

What do I need to do to prep Genexus Templating Strips 3B-GX5 and 4-GX5 (Cat. No. A49782) for use on the Genexus platform?

Remove the templating strips from their boxes in the refrigerator or freezer and get them ready them for loading in the sequencer by equilibrating to/thawing at room temperature for 30 mins. Confirm that the strip contents are completely thawed before loading in the sequencer. If you are delayed in loading, keep the thawed strips on ice or at 4 degrees C until you load them in the sequencer.

Genexus Strip 3B‑GX5 contain magnetic beads in one or two positions, yellow or brown in color, that sometimes get trapped in the upper “keyhole” of the tube. Dislodge these beads from the keyhole before installing the strip in the sequencer using the following procedure for each strip:
- Invert the strip 3-4 times to dislodge beads that are trapped in the keyholes.
- To remove any remaining beads and liquid from the keyholes, grasp the strip at one end with the strip seal facing up, then swing the strip with a rapid, downward centrifugal arm motion, ending with a sharp wrist-flick.
- Grasp the strip at the other end, then repeat the centrifugal motion.
- Check tube positions for significant amounts of beads that are still trapped in keyholes and then repeat the centrifugal motion, if needed. It is acceptable if a few beads remain in the keyhole or on the tube wall, but most should be either in suspension or in a pellet at the bottom of the tube.

What is the difference between Genexus Templating Strips 3B-GX5 and 4-GX5 (Cat. No. A49782) and Genexus Templating Strips 3-GX5 and 4 (Cat. No. A40263)?

Genexus Templating Strips 3B-GX5 and 4-GX5 (Cat. No. A49782) has new Genexus Strip 3B that has the chemistry modified to improve templating and sequencing of GC rich regions.

What application is Genexus Templating Strips 3B-GX5 and 4-GX5 (Cat. No. A49782) compatible with?

Genexus Templating Strips 3B-GX5 and 4-GX5 is specifically designed for use with the Oncomine Myeloid GX v2 assay. For all other Genexus assays, please use Genexus Templating Strips 3-GX5 and 4 (Cat. No. A40263).

What are the components of Genexus Templating Strips 3B-GX5 and 4-GX5 (Cat. No. A49782)?

Genexus Templating Strips 3B-GX5 and 4-GX5 provides 8 pairs of strips per kit - Genexus Templating Strips 3B-GX5 (Part No. A47712) (Orange, 8 strips) and Genexus Templating Strips 4-GX5 (Part No. A46816) (Yellow, 8 strips). Each pair of strips is required to perform automated template preparation and chip loading of two GX5 chips using the Genexus Integrated Sequencer.

What are the shipping and storage conditions for Genexus Templating Strips 3B-GX5 and 4-GX5 (Cat. No. A49782)?

Genexus Templating Strips 3B-GX5 (Part No. A47712) (Orange, 8 strips) ships on a gel pack and should be stored at 2 to 8 degrees C. Genexus Templating Strips 4-GX5 (Part No. A46816) (Yellow, 8 strips) ships on dry ice and should be stored at -30 to -10 degrees C.