The Amplitude High‐Throughput Consumable Package 2 Assay consists of assays and controls for 20,000 real-time RT-PCR tests for the detection of SARS-COV-2 RNA in nasal and nasopharyngeal swab samples. Laboratories can run up to 8,000 tests in 24 hours using the Amplitude Solution, and the consumable package format enables efficient ordering to address high-throughput needs.