Invitrogen™

Attune™ CytPix™ Violet 6 Conversion Kit, for yellow laser

Catalog number: A50929
Invitrogen™

Attune™ CytPix™ Violet 6 Conversion Kit, for yellow laser

Catalog number: A50929
Catalog Number
A50929
Unit Size
1 kit
Price (USD)
Catalog NumberUnit SizePrice (USD)
A509291 kit
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recommendations
The Attune CytPix Violet 6 Conversion Kit is designed to reconfigure Attune NxT four-channel violet instruments to allow detection of six channels of violet. A conversion kit is required for each original and any upgraded lasers of the instrument. The conversion is carried out by a Field Service Engineer.

Specifications

For Use With (Application)
Flow Cytometry
For Use With (Equipment)
Attune CytPix Flow Cytometer
Includes
Filters, dichroics
Product Line
DJM
Quantity
1 Kit
Regulatory Status
RUO
Shipping Condition
RT

Figures