Amphipol A8-35
Thermo Fisher Scientific™

Amphipol A8-35

Thermo Scientific Amphipol A8-35 is an amphipathic polymer ideally suited to improve solubilization and stabilization of membrane proteins while maintainingRead more
Have Questions?
Catalog NumberQuantity
A50943100 mg
Catalog number A50943
Price (MXN)
-
Quantity:
100 mg
Thermo Scientific Amphipol A8-35 is an amphipathic polymer ideally suited to improve solubilization and stabilization of membrane proteins while maintaining structural integrity and activity.

Features of Amphipol A8-35 include:
• Detergent-free substitute to extract and stabilize membrane proteins in aqueous solutions
• Surfactant with dense distribution of hydrophobic chains that have a high affinity for transmembrane surfaces to enable protein solubilization with minimal disruption of membrane environment
• Facilitates membrane protein structure/folding
• Suitable for protein solubilization and stabilization for single particle cryo-EM analysis

Properties of Amphipol A8-35:
Molecular formula: (C6.2H10.3O1.35N0.65Na0.35)∼72
Formula weight: ∼ 9 kDa
Solubility: up to 20% in water
Appearance: white solid

Citation(s):
1. Zoonens & Popot, J. Membr. Biol. 247:759-796, 2014.
2. Additional Amphipol references: www.ibpc.fr/amphipol

Specifications
ColorWhite
Formula Weight9 kDa
Molecular Formula(C6.2H10.3O1.35N0.65Na0.35)∼72
Solubility InformationUp to 20% in water
Physical FormSolid
Quantity100 mg
Unit SizeEach