AccelerOme™ Label-Free MS Sample Prep Kit, 1 x 16 Reactions - FAQs
AccelerOme™ Label-Free MS Sample Prep Kit, 1 x 16 Reactions - FAQs
View additional product information for AccelerOme™ Label-Free MS Sample Prep Kit - FAQs (A50944, A50945)
7 product FAQs found
Is the AccelerOme Label-Free MS Sample Prep Kit compatible with the BCA protein assay?
Yes, the buffer in the AccelerOme Label-Free MS Sample Prep Kit, in which samples are resuspended, is compatible with the BCA protein assay.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
Is the Lysis Buffer (Cat. No. A45735) compatible with the AccelerOme Label-Free MS Sample Prep Kit?
The Lysis Buffer (Cat. No. A45735) has been designed to be used with manual EasyPep MS Sample Prep Kits. The format of the Lysis Buffer container is not compatible with the AccelerOme platform and therefore it cannot be used with the AccelerOme Label-Free MS Sample Prep Kit.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
Can I use the samples obtained with the AccelerOme Label-Free MS Sample Prep Kit directly for mass spectrometry?
Yes, all buffers used in the AccelerOme Label-Free MS Sample Prep Kit are MS-compatible, and the samples can be directly used for mass spectrometry.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
Can the AccelerOme Label-Free MS Sample Prep Kit be reused?
The AccelerOme Label-Free MS Sample Prep Kit is a single-use kit, and it cannot be reused.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
Can I use the AccelerOme Label-Free MS Sample Prep Kit without the AccelerOme platform?
The AccelerOme Label-Free MS Sample Prep Kit has been developed to be used in the AccelerOme Automated Sample Preparation Platform. If you do not have the AccelerOme Automated Sample Preparation Platform, we recommend using EasyPep MS Sample Prep Kits (Cat. No. A40006, A57864, A45733 or A45734, depending on the number of reactions you would like to perform).
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
Which type of samples can be used with the AccelerOme Label-Free MS Sample Prep Kit?
The AccelerOme Label-Free MS Sample Prep Kit was initially optimized for proteins extracted from plasma, serum, cells, and tissue. However, it has also been demonstrated to work with bacteria and yeast samples. The AccelerOme Label-Free MS Sample Prep Kit allows working with samples containing between 1 to 100 µg of protein.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
How should I use the AccelerOme Label-Free MS Sample Prep Kit in the AccelerOme Automated Sample Preparation Platform?
All the buffers, reagents and other components in the AccelerOme Label-Free MS Sample Prep Kit come in a ready-to-use tray that has to be positioned into the AccelerOme Automated Sample Preparation Platform. Once placed on the device, the rest of the procedure is automated.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.