POROS™ GoPure™ Caprylate Pre-packed Column, 0.5 x 5 cm, 1 mL
Thermo Scientific™

POROS™ GoPure™ Caprylate Pre-packed Column, 0.5 x 5 cm, 1 mL

Thermo Scientific™ POROS™ GoPure™ pre-packed columns are intended for the screening of POROS chromatography resins or for small scale purification.
Have Questions?
Catalog NumberQuantity
A510541 column
Catalog number A51054
Price (EUR)
148,00
Each
Quantity:
1 column
Price (EUR)
148,00
Each

POROS GoPure pre-packed columns are intended for the screening of POROS chromatography resins or for small scale purification.

These pre-packed columns are ready to connect and have the following features:

  • Packed with POROS chromatography resins that offer superior performance over a wide range of flow rates
  • 1-mL and 5-mL formats provide a uniform screening platform for process development
  • Convenient, ready-to-run columns that simplify and speed up POROS resin evaluations and enable small scale purification
  • Direct connection to standard chromatography systems using 1/16 in., 10-32 fittings

POROS Caprylate Mixed-Mode Chromatography Resin (Cat. No. A51051) is designed to remove moderate to high levels of aggregates during downstream processing of therapeutic antibodies in flow-through mode. As a mixed-mode, weak cation-exchange resin, POROS Caprylate resin effectively removes other impurities, such as host cell proteins and a wide range of high molecular weight species.

Features of POROS Caprylate Mixed-Mode resin include:

  • Selective capture of aggregates and hydrophobic high-molecular-weight impurities
  • Flow-through design that helps save buffer and material costs
  • Excellent scalability and high productivity
  • Wide operational range
  • Non-animal-derived

POROS Caprylate Mixed-Mode resin operates in flow-through mode which helps reduce costs, especially when working with more engineered monoclonal antibody derivatives. The unique selectivity of the caprylate ligand contributes to a high yield of the targeted monomer while using established cation exchange resin buffer systems. POROS Caprylate Mixed-Mode resin is manufactured using a synthetic manufacturing process that is free of animal components.

Main characteristics

  • Matrix: cross-linked poly(styrene-divinylbenzene) POROS beads
  • Average particle size: 50 μm
  • Ligand: octanoic acid
  • Binding capacity: >20 g lysozyme/L resin
  • Typical buffers: typical cation exchange buffers, pH 4.5–7.0, conductivity 10–30 mS/cm
  • Clean-in-place: 0.5 M NaOH
  • Formulation buffer: 0.1 M NaOH
  • Mechanical resistance: 100 bar (10 MPa, 1450 psi)

Bulk resin is available and these columns come with a regulatory support package. Worldwide technical support is also available.

Other POROS chromatography resins are also available in this column format. All POROS high performance chromatography resins have a unique pore structure that provides excellent mass transport for high capacity and resolution over a wide range of flow rates.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Bed Volume1 mL
Column FormatChromatography Column
Particle Size50 μm
Quantity1 column
Stationary PhaseCaprylic Acid
TargetMonoclonal Antibodies
Product LinePOROS
TypeProcess Development
Unit SizeEach
Contents & Storage

Store at 2–30°C.