AAV-MAX Helper-Free AAV Production System Kit - FAQs
AAV-MAX Helper-Free AAV Production System Kit - FAQs
View additional product information for AAV-MAX Helper-Free AAV Production System Kit - FAQs (A51217)
13 product FAQs found
Are the components in the AAV-MAX Helper-Free AAV Production System available separately, or are they only sold as a kit?
The Viral Production Medium, Viral Production Cells 2.0, AAV-MAX Transfection Kit, Viral-Plex Complexation Buffer, and AAV-MAX Lysis Buffer are sold individually and here are the stand-alone catalog numbers:
- Viral Production Medium: Cat. No. A4817901, A4817902, A4817903
- Viral Production Cells 2.0: Cat. No. A49784, A51218
- AAV-MAX Transfection Kit: Cat. No. A50515, A50516
- Viral-Plex Complexation Buffer: A4983901
- AAV-MAX Lysis Buffer: A50520
Find additional tips, troubleshooting help, and resources within our Transfection Support Center.
Can I use other transfection reagents, such as PEI (polyethylenimine), in the AAV-MAX Helper-Free AAV Production System?
The AAV-MAX Transfection Reagent, AAV-MAX Tranfection Booster, and AAV-MAX Enhancer are specifically designed to work synergistically with the Viral Production Medium and Viral Production Cells 2.0 to provide maximum titers. Mixing and matching of reagents is not recommended, as this will decrease the overall performance.
Find additional tips, troubleshooting help, and resources within our Transfection Support Center.
How scalable is the AAV-MAX transfection protocol?
The AAV-MAX transfection protocol is scalable from 24 deep-well plates to shake flasks to bioreactors. The user guide provides guidance on this.
Find additional tips, troubleshooting help, and resources within our Transfection Support Center.
Are the LV-MAX Enhancer and AAV-MAX Enhancer different?
Yes, these are different, and each is optimized for its intended use.
Find additional tips, troubleshooting help, and resources within our Transfection Support Center.
Will I see a drop in adeno-associated virus (AAV) titer if I don't use the AAV-MAX Enhancer with the AAV-MAX Helper-Free AAV Production System?
The AAV-MAX Enhancer is important for boosting AAV production. If the enhancer is not used, it will reduce system performance by 50% or more.
Find additional tips, troubleshooting help, and resources within our Transfection Support Center.
What is the optimal incubation time for the plasmid DNA/AAV-MAX Transfection Reagent/AAV-MAX Transfection Booster complex?
We recommend a complexation time of 20-30 mins. We recommend optimizing the complexation time for different sized plasmids.
Find additional tips, troubleshooting help, and resources within our Transfection Support Center.
Is it necessary to use the AAV-MAX Transfection Booster with the AAV-MAX Helper-Free AAV Production System? Will I see a drop in adeno-associated virus (AAV) titer if the booster is not used?
Yes, the AAV-MAX Transfection Booster facilitates transfection. If the booster is not used, it will reduce system performance.
Find additional tips, troubleshooting help, and resources within our Transfection Support Center.
Do I need to supplement the Viral Production Medium prior to use?
Yes, prior to using the Viral Production Medium, it should be supplemented with 4 mM GlutaMAX Supplement.
Can I use Viral Production Cells (Cat. No. A35347, A35827) with the AAV MAX Helper-Free AAV Production System?
The Viral Production Cells 2.0 are specifically designed to work with the other components of the AAV MAX Helper-Free AAV Production System to provide maximal adeno-associated virus (AAV) titers. Viral Production Cells could be used but we don't recommend doing this as it will reduce system performance.
What is the highest density that Viral Production Cells 2.0 can be cultured at?
The Viral Production Cells 2.0 achieve maximum cell densities of >1.2 x 10E7 cells/mL in shake flask cultures.
What is the passage stability of the Viral Production Cells 2.0?
We recommend using the Viral Production Cells 2.0 between passages 3 to 25 for adeno-associated virus (AAV) production.
How long do the Viral Production Cells 2.0 require to recover post-thaw before transfection?
We recommend allowing freshly thawed Viral Production Cells 2.0 to recover in culture for at least three passages prior to transfection.
Are the Viral Production Cells 2.0 a clonal cell line?
Yes, the Viral Production Cells 2.0 cells are a clonal cell line derived from single cell cloning of 293-F cells.