The Thermo Scientific VitroEase Apoferritin Standard is a highly validated pure protein for use as a quality control sample forRead more
Have Questions?
Catalog Number
Quantity
A51362
5 x 20 μL
Catalog number A51362
Price (KRW)
761,000
Online offer
Ends: 30-Sep-2026
869,000
Save 108,000 (12%)
Each
Quantity:
5 x 20 μL
Price (KRW)
761,000
Online offer
Ends: 30-Sep-2026
869,000
Save 108,000 (12%)
Each
The Thermo Scientific VitroEase Apoferritin Standard is a highly validated pure protein for use as a quality control sample for cryo-electron microscopy single particle analysis.
Features of the VitroEase Apoferritin Standard include: • Positive control sample—apoferritin protein production optimized and validated for use in cryo-EM grid preparation and analysis • Rigorously tested—high-quality, consistent protein standard documented via lot-specific certificates of analysis • Convenient—ready-to-use format with each single-use vial sufficient for 5–6 grid preparations
Protein sample grid preparation is a major bottleneck in cryo-electron microscopy (cryo-EM) single particle analysis (SPA), often requiring days or even weeks to optimize. The VitroEase Apoferritin Standard has been designed specifically to meet the stringent needs for high protein purity, minimal protein aggregation, and intact protein structure. Apoferritin, a 24-subunit protein (474 kDa), is considered the gold standard for cryo-EM SPA, and this standard is fit for use as a comparison for success for new users or as quality control for expert users.
Specifications
ColorColorless
Concentration3.5–4.0 mg/mL
Detection MethodCryo-EM
Expression SystemBacterial
For Use With (Equipment)Cryo-Electron Microscopes, Mass spectrometers
Gene AliasApoF
Gene SymbolFTH1, FTH
Ligand TypeIron
NameApoferrtin
Quantity5 x 20 μL
Sample TypePurified protein
SterilityNon-sterile
TechniqueMS, Cryo-EM SPA
Volume (Metric)100 μL
For Use With (Application)Cryo-EM Single Particle Analysis (SPA)
FormLiquid
Unit SizeEach
Frequently asked questions (FAQs)
Is VitroEase Apoferritin Standard suitable for high resolution cryo-EM (electron microscopy)?
VitroEase Apoferritin Standard is suitable for high resolution cryo-EM (electron microscopy) and we have imaged it at 1.8 angstroms using our Krios G4 Cryo-TEM instrument.
What is the origin species for VitroEase Apoferritin Standard?
VitroEase Apoferritin Standard is human apoferritin heavy chain expressed in E. coli.