MicroSEQ™ ID Software for SeqStudio™ Genetic Analyzer v1.0, with all library kits - FAQs
MicroSEQ™ ID Software for SeqStudio™ Genetic Analyzer v1.0, with all library kits - FAQs
View additional product information for MicroSEQ™ ID Software for SeqStudio™ Genetic Analyzer v1.0, with all library kits - FAQs (A51427)
11 product FAQs found
Which Thermo Fisher Scientific instrument laptops are compatible with MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0 (Lite Installation)?
The Dell Latitude 3541 Laptop with WIN10 IOT 2018-64-bit is the Thermo Fisher Scientific instrument laptop that is compatible with MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0 (Lite Installation).
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Which Thermo Fisher Scientific instrument computers are compatible with MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0 (System and Lite Installations)?
The Dell Optiplex XE3 with WIN10 IOT 2018-64-bit is the Thermo Fisher Scientific instrument computer that is compatible with MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0 (System and Lite Installations).
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Which KB Basecaller version is supported for MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0?
KB Basecaller v1.4.2.5 is supported for MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What are the computer requirements for MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0 Lite Installation?
Here are the computer requirements for MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0 Lite Installation:
- WIN 10 IoT 64-bit
- Intel Core i5 Processor, 2.4GHz or faster, 16GB RAM, and at least 250 GB for software installation and data storage
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What are the computer requirements for MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0 System Installation?
Here are the computer requirements for MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0 System Installation:
- WIN 10 IoT 64-bit
- Intel Core i7 Processor, 3.1GHz or faster, 16GB RAM, and at least 250 GB for software installation and data storage
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What Windows OS is MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0 compatible with?
MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0 is compatible with 64-bit Windows 10 OS.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Which MSID Library versions is MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0 compatible with?
MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0 is compatible with the following MSID libraries:
- MSID 16S rDNA Full Gene Library v2.0
- MSID Fungal Library v2018
- MicroSeq_ID_16S_rDNA_500_Library_v2019
- MicroSEQ_ID_16S_rDNA_500_Supplemental_Library_v2019
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Which SeqStudio Genetic Analyzer SAE Admin Console version is MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0 compatible with?
MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0 is compatible with SeqStudio Genetic Analyzer SAE Admin Console v2.0.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Which SeqStudio Genetic Analyzer firmware version is MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0 compatible with?
MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0 is compatible with SeqStudio Genetic Analyzer firmware version 1.2.2.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Which Data Collection Software version is MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0 compatible with?
MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0 is compatible with SeqStudio Data Collection Software v1.2.1 and v1.2.3.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What is the catalog number for MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0, with all library kits?
The catalog number for MicroSEQ ID Software for SeqStudio Genetic Analyzer v1.0, with all library kits is A51427.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.