Pierce™ FlexMix™ Calibration Solution for Auto-Ready Mass Spectrometers - FAQs
Pierce™ FlexMix™ Calibration Solution for Auto-Ready Mass Spectrometers - FAQs
View additional product information for Pierce™ FlexMix™ Calibration Solution for Auto-Ready Mass Spectrometers - FAQs (A51739)
4 product FAQs found
In which mass spectrometer instruments can I use the Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers?
Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers is designed to be used in mass spectrometer instruments with autocalibration feature, such as Orbitrap IQ-X Mass Spectrometer using Orbitrap Tribrid Series 3.5 Instrument Control Software (or later).
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
Does the Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers need to be diluted?
Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers is a ready to use reagent and there is no need for dilution.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
Is there a specific mode in which Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers needs to be run?
Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers can be run in Positive or Negative mode.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
What is the difference between Pierce FlexMix Calibration Solution and Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers?
Both the Pierce FlexMix Calibration Solution and Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers have the same 16 components and produce the same spectra. However, the Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers has a different storage solution: LC-MS grade acetonitrilie 87% (v/v) and LC-MS grade water 13% (v/v). The FlexMix Calibration for Auto-Ready Mass Spectrometers is designed to be used only in mass spectrometer instruments with autocalibration feature, such as Orbitrap IQ-X Mass Spectrometer using Orbitrap Tribrid Series 3.5 Instrument Control Software (or later).
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.