Pierce™ Small Molecule System Suitability Standard
Thermo Scientific™

Pierce™ Small Molecule System Suitability Standard

The Thermo Scientific Pierce Small Molecule System Suitability Standard provides a pre-formulated mixture of nine small-molecule standards to assess systemRead more
Have Questions?
Catalog NumberQuantity
A5174010 injections, 0.25 mL
Catalog number A51740
Price (USD)
495.00
Each
Quantity:
10 injections, 0.25 mL
Price (USD)
495.00
Each
The Thermo Scientific Pierce Small Molecule System Suitability Standard provides a pre-formulated mixture of nine small-molecule standards to assess system performance in both positive and negative ionization modes for TSQ Triple Quadrupole and Orbitrap Exploris mass spectrometers.

Pierce Small Molecule System Suitability Standard features include:
• Comprehensive—mixture of nine small-molecule compounds (mass range: 76–1279) designed to assess instrument performance in both positive and negative ionization modes
• Highly validated—high-quality, rigorously-tested formulation with lot-specific certificates of analysis
• Convenient—pre-formulated ready-to-use liquid format
• Diagnostic—excellent QC assay tool for quality assessment of LC and MS instrument status

The Pierce Small Molecule Suitability Standard (SMSS) is a ready-to-use formulation optimized for the assessment of MS instrument performance for metabolomics applications. The standard contains a mixture of nine compounds with a mass range of 76–1279, appropriate for small molecule analysis (see table below). The standard has been designed for use with TSQ Triple Quadrupole and Orbitrap Exploris MS series instruments to verify individual system and multiple system performance. Additionally, the SMSS enables quicker identification of sub-optimal system performance, enhances troubleshooting, and provides standardization to help obtain higher quality data. The SMSS is manufactured at an ISO 9001 facility and each lot is quality controlled with strict specifications.

Specifications
ColorColorless
GradeMS Grade
Physical FormLiquid
Quantity10 injections, 0.25 mL
Unit SizeEach

Frequently asked questions (FAQs)

In which mode can I run the Small Molecule Suitability Standard?

The Small Molecule Suitability Standard can be run in Positive or Negative mode.

Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.

Does the Small Molecule Suitability Standard need to be diluted?

The Small Molecule Suitability Standard can be diluted 1:10 (25 µL of standard with 225 µL of 0.1% Formic Acid) or can be used without dilution. Glass vials are provided for dilutions. It is important to use 0.1% Formic Acid solution for dilution, in order to keep all components in solution.

Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.

What is the mass range for the Small Molecule Suitability Standard?

The mass range for the Small Molecule Suitability Standard (SMSS) is 76 to 1279 m/z.

Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.