Search
Search

| 카탈로그 번호 | 반응 수 |
|---|---|
| A51803 | 1000 Reactions |
| A51801 | 100 Reactions |
| A51802 | 400 Reactions |
Invitrogen SuperScript IV RT-LAMP Master Mix is a ready-to-use solution optimized for fast, sensitive, and simple RNA target detection in loop-mediated isothermal amplification (LAMP) reactions. With this convenient master mix formulation, you can perform isothermal amplification from both RNA or DNA targets in a single tube format.
Benefits of SuperScript IV RT-LAMP Master Mix
SuperScript IV RT-LAMP Master Mix includes an evolved Bst DNA polymerase mutant with a fast reaction speed and increased sensitivity and tolerance to inhibitors, as well as SuperScript IV Reverse Transcriptase with increased processivity, thermostability, and inhibitor resistance. To prevent RNA degradation by carry-over RNases, the master mix includes the potent, noncompetitive RNaseOUT Recombinant Ribonuclease Inhibitor. Our proprietary reaction buffer composition is optimized to increase specificity by delaying no-template control (NTC) amplification time.
• 2X SuperScript IV RT-LAMP Master Mix (10 x 1.25 mL)
• SYTO Green Fluorescent Nucleic Acid Stain (30 μL)
Store at –20°C and protect from light.