Product Image
Applied Biosystems™

TaqMan™ Antibiotic Resistance Assay, pooled

Applied Biosystems TaqMan Antibiotic Resistant assays are used for quantitative real-time PCR detection of microbial species. These assays consist ofRead more
Have Questions?
Catalog NumberQuantity
A52079250 μL
Catalog number A52079
Price (USD)
344.00
Each
Quantity:
250 μL

Applied Biosystems TaqMan Antibiotic Resistant assays are used for quantitative real-time PCR detection of microbial species. These assays consist of up to four pooled unlabeled PCR primers and TaqMan probes with a dye label (FAM) on the 5' end and a minor groove binder (MGB) and non-fluorescent quencher (NFQ) on the 3' end.

Features of TaqMan Antibiotic Resistance assays include:

  • Easy to use—just add DNA and master mix and run the qPCR—no melt curves required
  • Specific—TaqMan assays use proprietary MGB-containing probes that can be up to 15 bases shorter than non-MGB probes, improving the specificity of the assay
  • Sensitive—TaqMan assays are ideal for measuring low-abundance microbial targets
  • Accurate—identify microbial targets with high accuracy of quantitation
  • Extensive content—over 130 pre-designed antibiotic resistance assays currently available
  • Gold-standard TaqMan qPCR chemistry—TaqMan assays draw on Thermo Fisher Scientific's bioinformatics assay design pipeline to help ensure high specificity and minimal cross-reactivity, even for closely related microbial species and subtypes
  • Fast delivery—assays shipped in as little as 1–2 days (see below)

Approximate ship time

  • Inventoried: 1–2 days in North America*
  • Made to order: 4–6 days in North America

All of our predesigned TaqMan gene expression assays are covered by the TaqMan Assays qPCR Guarantee.**

*Our most popular assays are stocked in North America for overnight delivery where possible. If an assay is out of stock, it may take an additional 3–5 business days for delivery.

**Subject to terms and conditions. For complete details, go to www.thermofisher.com/taqmanguarantee.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
3'Primer ModificationFAM
5'Primer ModificationMGB-NFQ
FormatTube
No. of Reactions250 Reactions
Product LineTaqMan
Quantity250 μL
Shipping ConditionDry Ice
Concentration20X
Detection MethodPrimer-probe
For Use With (Application)Pathogen Detection
PCR MethodqPCR
Unit SizeEach
Contents & Storage
Store at -15 to -25°C.