Can I use Efficient-Pro Feed 2 with other cell lines besides CHO-S and DG-44 cell line variants?
Efficient-Pro Feed 2 has been designed specifically for CHO-S and DG-44 cells; however, it has been demonstrated to improve performance in other CHO variants and even other parental cell lines.