Mass Spectrometry Compatible Crosslinkers
Thermo Scientific™

Mass Spectrometry Compatible Crosslinkers

Improve mass spectrometry detection of crosslinked proteins and interactions with DSPP and TBDSPP heterotrifunctional crosslinkers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
A52286DSPP
A52287TBDSPP
Catalog number A52286
Price (USD)
540.60
Special offer
Online exclusive
Ends: 15-Jul-2026
636.00
Save 95.40 (15%)
Each
Description:
DSPP
Price (USD)
540.60
Special offer
Online exclusive
Ends: 15-Jul-2026
636.00
Save 95.40 (15%)
Each
Chemical crosslinking in combination with mass spectrometry is a powerful method to improve protein characterization, and to identify protein-protein interactions. This method has been applied to recombinant and native protein complexes, and more recently, to whole cell lysates or intact unicellular organisms in efforts to identify protein-protein interactions on a global scale. The enrichment feature of DSPP and TBDSPP improves detection of crosslinked proteins by reducing sample complexity through separation of crosslinked from non-crosslinked peptides.
• Tri-functional crosslinker: NHS ester (both ends), phosphonic acid (DSPP), or phosphonate ester (TBDSPP) in the spacer for enrichment
• High-purity crystalline reagents for protein structure and interaction characterization
• Membrane-permeable version (TBDSPP) for intracellular crosslinking
• Enrichable using Fe-NTA IMAC or TiO2 MOAC

DSPP (disuccinimidyl phenyl phosphonic acid, PhoX) and TBDSPP (tert-butyl disuccinimidyl phenyl phosphonate, tBu-PhoX) are phospho-enrichable crosslinkers containing an amine-reactive N-hydroxysuccinimide (NHS) ester at each end of a 7-atom spacer arm with a phosphonic acid group (DSPP) or phosphonate ester (TBDSPP) for enrichment. Both react efficiently with primary amine groups (-NH2) in pH 7–9 buffers to form stable amide bonds. The phosphonic acid or phosphonate ester groups can then be used to enrich crosslinked peptides using immobilized metal affinity chromatography (IMAC) or metal oxide affinity chromatography (MOAC).

Of the two enrichable MS crosslinkers, DSPP is more water soluble than TBDSPP and is better for in vitro crosslinking, simple purified proteins and protein complexes. TBDSPP is better for intracellular crosslinking due to increased membrane permeability. Although both crosslinkers can be enriched using traditional phosphopeptide methods, TBDSPP crosslinked peptides must first be incubated with TFA to remove the tert-butyl protection groups. The TBDSPP phosphonic acid group (after acid cleavage) is not a substrate for phosphatases, which provides opportunity for the removal endogenous phosphopeptides in samples prior to enrichment.

Note: For high resolution analysis of the crosslinked peptides, the recommended LC column for the Nanospray Flex source is the Acclaim PepMap 100 C18 LC Column (Cat. No. 164940 or 164568). For the EASY-Spray source, the recommended LC column is the EASY-Spray C18 LC Column (Cat. No. ES900 or ES904).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell PermeabilityNo
DescriptionDSPP
FormSolid
Labeling MethodChemical Labeling
Molecular Weight (g/mol)440.25
PEGylatedNo
Product LineMS Reagents
Purity90%
Quantity50 mg
Reactive MoietyNHS Ester
SolubilityWater
Spacer Arm Length4.3 Å
Water SolubleYes
Chemical ReactivityAmine-Amine
CleavableNo
Crosslinker TypeHomotrifunctional
FormatStandard
Product TypeCrosslinker
SpacerMedium (10 to 30 Å)
Unit SizeEach
Contents & Storage
Upon receipt store at -20°C protected from moisture.

Frequently asked questions (FAQs)

What is the advantage of using phospho-enrichable crosslinkers?

Phospho-enrichable crosslinkers such as DSPP (Disuccinimidyl Phenyl Phosphonic Acid, PhoX) and TBDSPP (tert-Butyl Disuccinimidyl Phenyl Phosphonate, tBu-PhoX) are great tools for studying protein-protein Interaction, protein 3D structure, and for mass spectrometry (MS) analysis. They can be used to enrich low-abundance crosslinked peptides and thereby improve MS identification rates.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Is DSPP (Disuccinimidyl Phenyl Phosphonic Acid, PhoX) cell-permeable?

DSPP is not cell-permeable. We offer TBDSPP (tert-Butyl Disuccinimidyl Phenyl Phosphonate, tBu-PhoX), Cat. No. A52287 that is a cell-permeable phopho-enrichable crosslinker.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.