CTS™ DynaCellect™ Cell Isolation Kit
Gibco™

CTS™ DynaCellect™ Cell Isolation Kit

The Gibco CTS DynaCellect Cell Isolation Kit is a single-use, fit-for-purpose kit designed for use with the CTS DynaCellect MagneticRead more
Have Questions?
Catalog NumberQuantity
A523005 kits
Catalog number A52300
Price (CLP)
-
Quantity:
5 kits

The Gibco CTS DynaCellect Cell Isolation Kit is a single-use, fit-for-purpose kit designed for use with the CTS DynaCellect Magnetic Separation System, which is an automated instrument for T-cell isolation and activation, as well as magnetic bead removal. This kit is used for isolation and activation of immune cells in cell processing applications.

Characteristics of the CTS DynaCellect Cell Isolation Kit include:

  • Weldable tube material: 8 x DEHP-free PVC
  • Working volume size: 10 mL–1 L
  • Bag chamber size: 1 L

This kit and the CTS DynaCellect Bead Removal Kit enable a range of cell processing applications such as isolation, depletion, and bead removal. They offer flexibility for process development and can optimized to support commercial scale manufacture.

Each kit has input/output sterile weldable tubes for flexible setup configurations. They are compatible with sterile welding and are gamma sterilized.

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.
Specifications
Capacity (Metric)1 L
Quantity5 kits
Product TypeIsolation Kit
Unit SizeEach