Fetal Bovine Serum, Value
Gibco™

Fetal Bovine Serum, Value

Los sueros fetales bovinos Gibco ofrecen un excelente valor para el cultivo celular básico, la investigación especializada y los ensayos específicos. Se ha ganado la confianza de los investigadores con una calidad uniforme y un soporte galardonado que ayuda a satisfacer las necesidades de investigación y presupuesto.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidadFormatoInactivado por calorGamma-irradiated
A5256501500 mLFrascoNoSí
A5256701500 mLFrascoNoNo
A5256901500 mLFrascoSíSí
Número de catálogo A5256501
Precio (MXN)
13,589.15
Each
Cantidad:
500 mL
Formato:
Frasco
Inactivado por calor:
No
Gamma-irradiated:
Sí
Precio (MXN)
13,589.15
Each

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

Sera category: Value

  • Use for general cell culture with common cell lines
  • Low endotoxin and hemoglobin levels
  • Endotoxin level: <20 EU/mL
  • Hemoglobin level: <25 mg/dL
  • Origin: refer to CoA for specific origin. All products may not be available in all regions due to importation regulations.
  • US Department of Agriculture (USDA) safety tested for the absence of bluetongue and akabane from countries that are USDA-approved for importation into the US

Gibco serum delivers

  • ISO 13485–certified; processed in FDA registered facilities
  • Triple-filtered at 0.1 μm
  • Gibco bottle easier to use in hood; helps reduce risk of contamination and improve consistency
  • Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook

* FBS import requirements vary globally, and acceptance depends on whether competent authorities allow import from a specific country of origin. Please check applicable regulations for the export of FBS and any FBS-containing products (e.g., cultured cells). Contact your Thermo Fisher Scientific representative for support.

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.
Especificaciones
Concentración de endotoxinas≤20 EU/mL
Para utilizar con (aplicación)*FBS import requirements vary globally, and acceptance depends on whether competent authorities allow import from a specific country of origin. Please check applicable regulations for the export of FBS and any FBS-containing products (e.g., cultured cells). Contact your Thermo Fisher Scientific representative for support.
Gamma-irradiatedSí
GradoValue
Inactivado por calorNo
Concentración de hemoglobina≤25 mg/dL
Sustancias contaminantesTriple filtered at 0.1 μm
Línea de productos0Y7
Grado de pureza o calidadValue
Duración de almacenamiento5 Years
Condiciones de envíoDry Ice
EspecieCattle/Bovine
Pruebas de seguridad del USDA incluidasUSDA Safety Testing Performed for Australian origins
Volumen (métrico)500 mL
AgeFetal
País de origenConsulte CoA para conocer el origen específico*
FormularioFrozen
FormatoFrasco
Tipo de productoFetal Bovine Serum
Cantidad500 mL
Serum TreatmentStandard (Sterile-filtered)
EsterilidadSterile
Sterilization MethodTriple-filtered, 0.1 μm
Unit SizeEach
Contenido y almacenamiento
Storage conditions: ≤-10°C