Fetal Bovine Serum, Value
Gibco™

Fetal Bovine Serum, Value

Gibco fetal bovine sera offer an excellent value for basic cell culture, specialty research, and specific assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormatHeat InactivatedGamma-irradiated
A5256901500 mLBottleYesYes
A5256501500 mLBottleNoYes
A5256701500 mLBottleNoNo
Catalog number A5256901
Price (MXN)
14,521.74
Each
Quantity:
500 mL
Format:
Bottle
Heat Inactivated:
Yes
Gamma-irradiated:
Yes
Price (MXN)
14,521.74
Each

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

Sera category: Value

  • Use for general cell culture with common cell lines
  • Low endotoxin and hemoglobin levels
  • Endotoxin level: <20 EU/mL
  • Hemoglobin level: <25 mg/dL
  • Origin: refer to CoA for specific origin. All products may not be available in all regions due to importation regulations.
  • US Department of Agriculture (USDA) safety tested for the absence of bluetongue and akabane from countries that are USDA-approved for importation into the US

Gibco serum delivers

  • ISO 13485–certified; processed in FDA registered facilities
  • Triple-filtered at 0.1 μm
  • Gibco bottle easier to use in hood; helps reduce risk of contamination and improve consistency
  • Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook

* FBS import requirements vary globally, and acceptance depends on whether competent authorities allow import from a specific country of origin. Please check applicable regulations for the export of FBS and any FBS-containing products (e.g., cultured cells). Contact your Thermo Fisher Scientific representative for support.

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.
Specifications
Endotoxin Concentration≤20 EU/mL
For Use With (Application)*FBS import requirements vary globally, and acceptance depends on whether competent authorities allow import from a specific country of origin. Please check applicable regulations for the export of FBS and any FBS-containing products (e.g., cultured cells). Contact your Thermo Fisher Scientific representative for support.
Gamma-irradiatedYes
GradeValue
Heat InactivatedYes
Hemoglobin Concentration≤25 mg/dL
ProcessingTriple filtered at 0.1 μm
Product Line0Y7
Purity or Quality GradeValue
Shelf Life5 Years
Shipping ConditionDry Ice
SpeciesCattle/Bovine
USDA Safety Testing IncludedUSDA Safety Testing Performed for Australian origins
Volume (Metric)500 mL
AgeFetal
Country of OriginRefer to CoA for specific origin*
FormFrozen
FormatBottle
Product TypeFetal Bovine Serum
Quantity500 mL
Serum TreatmentStandard (Sterile-filtered)
SterilitySterile
Sterilization MethodTriple-filtered, 0.1 μm
Unit SizeEach
Contents & Storage
Storage conditions: ≤-10°C