VetMAX™ PRRSV 3.0 Reagents
Applied Biosystems™

VetMAX™ PRRSV 3.0 Reagents

VetMAX™ PRRSV 3.0 Reagents contain master mix, Xeno RNA control, and a primers/probes mix that enables multiplex, one-step, reverse transcription and real-time PCR (RT-PCR) assays targeting North American (NA) and European (EU) porcine reproductive and respiratory syndrome virus (PRSSV) RNA and Xeno RNA.
Have Questions?
Catalog NumberQuantity
A52576100 Reactions
Catalog number A52576
Price (MXN)
-
Quantity:
100 Reactions

VetMAX PRRSV 3.0 Reagents contain master mix, Xeno RNA control, and a primers/probes mix that enable multiplex, one-step, reverse transcription and real-time PCR (RT-PCR) assays targeting North American (NA) and European (EU) porcine reproductive and respiratory syndrome virus (PRSSV) RNA and Xeno RNA. The Xeno RNA control (10,000 copies/μL) is added during extraction as an internal positive control (IPC). These reagents contain an updated primer/probe mix compared to the similar VetMAX NA and EU PRRSV 2.0 Reagents.

Features of the VetMAX PRRSV 3.0 Reagents include:

  • Rapid and cost-effective method for amplification of target sequences
  • Quality reagents manufactured under ISO 9001, providing confidence in supply and quality
  • From a brand with over 20 years in the DNA/RNA arena, with extensive bioinformatics capabilities and experience

PRSSV causes significant losses in the swine industry due to reproductive disorders, respiratory conditions, and growth retardation. PRRSV can be identified in blood and semen. Real-time PCR is the most sensitive method for analyzing RNA amplification.

For Research Use Only
Specifications
DiseasePorcine Reproductive and Respiratory Syndrome Virus
No. of Reactions100 Reactions
Quantity100 Reactions
Sample TypeSerum, Oral Fluids, Tissue, Semen, Processing Fluid, Whole Blood
Shelf Life24 Months
Shipping ConditionShipped at -30°C to -10°C
Concentration2X (Master Mix), 10X (Primer/Probe Mix)
Unit SizeEach
Contents & Storage
Store at -10° to -30°C.