Genexus™ Purification System Extended Warranty, Qualification & Training Package
Ion Torrent™
Genexus™ Purification System Extended Warranty, Qualification & Training Package
The Ion Torrent™ Genexus™ Purification System automates the extraction of nucleic acids in as little as two hours from various sample types. This package includes the Genexus Purification System, SmartStart Orientation, Qualification Services, and a one- or two-year extended warranty.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
A525811 paquete/2 años de garantía ampliada
A525801 paquete/1 año de garantía ampliada
Número de catálogo A52581
Precio (CLP)
-
Cantidad:
1 paquete/2 años de garantía ampliada
The Ion Torrent Genexus Purification System automates the extraction of nucleic acids in as little as two hours from various sample types, including formalin-fixed, paraffin-embedded (FFPE) lysate, plasma, whole blood, fresh or frozen tissue, peripheral blood leukocytes (PBLs), cultured cells, and bone marrow aspirate. Consumables for the system come prefilled with reagents and support up to 12 samples per run (6 samples when extracting from plasma). The system includes an onboard Qubit Fluorometer to automate post-extraction nucleic acid quantification and has been verified using the Genexus Integrated Sequencer as part of a complete specimen-to-result workflow.
This package includes the Genexus Purification System, SmartStart Orientation, qualification services, and a one- or two-year extended warranty.
The Genexus Purification System Package offers:
MagMAX technology—enabling consistent extraction results
Prefilled consumables—helping reduce setup hands-on time
Vision system—providing real-time feedback on consumables during run setup to prevent setup error
Integrated nucleic acid quantitation—reducing post-run processing times
• 1 day SmartStart Orientation to get you up and running quickly
• Qualification services to be provided at installation and after every planned maintenance or critical repair to ensure your system maintains regulatory compliance
• AB Assurance extended warranty including comprehensive service coverage and one planned maintenance visit
The Genexus Purification System automates the extraction and quantitation of nucleic acids from a variety of tissue types for use on the Genexus Integrated Sequencer. With a single touch point and 10 minutes of hands-on time, the purification system can extract both DNA and RNA sequentially from FFPE lysates, cfTNA from plasma, and DNA or RNA separately from whole blood, fresh or frozen tissue, bone marrow, or peripheral blood leukocytes. When connected to the integrated sequencer, the Genexus Software streamlines the next-generation sequencing (NGS) workflow by integrating the setup-to-report workflow within a single software ecosystem.
Prefilled reagents
The Genexus Purification System consumables are prefilled with most reagents necessary to perform the extraction, so only a few pipetting steps are required per sample to set up the run. Simply remove the foil seal, add enzymes and your samples, then place the consumables onto the instrument.
Vision system
Just like the Genexus Integrated Sequencer, the Genexus Purification System uses an onboard vision system to verify reagent placement and detect errors in real time. The vision system continuously checks that the consumables are appropriate to the purification run type and helps prevent the running of expired or previously used consumables. Feedback on the consumables loaded in the system is displayed on the screen in the system’s base.
MagMAX technology
MagMAX technology uses magnetic beads to perform nucleic acid extraction. MagMAX beads offer superior binding capacity due to their large surface area, accept a greater range of samples with differing viscosities, and allow for efficient elution in small volumes.
48‐well nucleic acid archive plate
After extraction, all nucleic acids are transferred directly to the 48‐well archive plate and kept cool for up to one hour after run completion. The 48‐well nucleic acid archive plate is barcoded to facilitate the tracking of sample extractions across several stored archive plates.
Automated quantitation
The Genexus Purification System automates quantitation of extracted nucleic acids using an onboard Qubit Fluorometer. Two dyes are used to measure the concentration of DNA or RNA from each extracted sample. Automating post-run quantitation saves hands-on time and allows for immediate use of the extracted samples in genetic analysis workflows after run completion.
Rapid turnaround
Run times on the Genexus Purification System range from 2 to 5.5 hours depending on the number of samples extracted and the protocol being run. Automated quantitation helps significantly reduce post-run hands-on time, allowing for rapid transition to subsequent genetic analysis workflows.
Post-run clean
After each run all consumables that touch the sample are discarded by the researcher after which the Genexus Purification System uses a UV light to clean the deck. This automated UV cleaning prevents possible run-to-run carryover of nucleic acid which could lead to contamination of downstream sequencing.
Get up and running fast with SmartStart Orientation
The SmartStart Orientation included in this package is performed by a Field Application Scientist (FAS) remotely or in your lab and helps you:
• Get up and running quickly
• Get connected to your local network and Cloud
• Choose appropriate reagents and consumables to meet your needs
Content will vary depending on your level of knowledge and experience.
Qualification services help ensure regulatory compliance
This package includes Installation, operational and instrument performance verification (IQOQ/IPV) services. IQOQ documents that the instrument has been installed and operates according to manufacturer’s specifications. IPV documents that the system can perform effectively and reproducibly within performance specifications. IQOQ and IPV are recommended at installation, after moving the instrument, upgrading software or hardware, planned maintenance or critical repairs.
Service plans help protect your investment
Service plans such as the AB Assurance extended warranty included in this package help to:
• Maximize system uptime with fast repair turnaround time from manufacturer-trained and certified field service engineers (FSEs)
• Reduce overall repair costs with remote instrument diagnosis and resolution
• Extend the life of your instrument and help keep it running at peak performance with annual maintenance service
During the annual maintenance, the FSE will update software if required, clean, lubricate, and align relevant parts and optical systems, functional tests as well as document tasks, results, and findings.
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Customer EducationSmart Start 1 day at customer site
Tipo de producto finalDNA/RNA from FFPE Lysate, DNA/RNA from Blood, PBLs, Bone Marrow Lysate, Tissue Homogenate, or cfTNA from Plasma
IncluyeAutomated Quantitation
Tecnología de aislamientoMagMax
Cantidad1 paquete/2 años de garantía ampliada
Muestras por serie12 (6 for Plasma)
Total Service CoverageIQOQ at installation, 1 year manufacturer warranty, and 2 year of field service with PM with OQPQ
Unit SizeEach