What is the alternative for Absolute Q AAV dPCR Assay (Cat. No. A52740)?
We recommend the Absolute Q Viral Titer Assays for digital PCR (Cat. No. A53736) as an alternative to Cat. No. A52740.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.