Slide-A-Lyzer™ G3 Dialysis Cassettes, 10K MWCO
Slide-A-Lyzer™ G3 Dialysis Cassettes, 10K MWCO
Slide-A-Lyzer™ G3 Dialysis Cassettes, 10K MWCO
Slide-A-Lyzer™ G3 Dialysis Cassettes, 10K MWCO
Slide-A-Lyzer™ G3 Dialysis Cassettes, 10K MWCO
Slide-A-Lyzer™ G3 Dialysis Cassettes, 10K MWCO
Thermo Scientific™

Slide-A-Lyzer™ G3 Dialysis Cassettes, 10K MWCO

Thermo Scientific Slide-A-Lyzer™ G3 Dialysis Cassettes facilitate the rapid and trouble-free dialysis of sample volumes from 0.5 to 125 mL. Unlike standard flat tubing, these innovative devices do not require knots or clips that can lead to leaking and sample loss.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Quantity
A529713 mL10 cassettes
A5297215 mL8 cassettes
A5297330 mL6 cassettes
A5297470 mL6 cassettes
A52975125 mL6 cassettes
Catalog number A52971
Price (USD)
150.65
Online Exclusive
165.00
Save 14.35 (9%)
Each
Capacity (Metric):
3 mL
Quantity:
10 cassettes
Price (USD)
150.65
Online Exclusive
165.00
Save 14.35 (9%)
Each

Thermo Scientific Slide-A-Lyzer G3 Dialysis Cassettes facilitate the rapid and trouble-free dialysis of sample volumes from 1 to 125 mL.They are single-use dialysis devices designed to maximize performance and practicality. They provide an easy-to-use and secure alternative to traditional dialysis tubing and are used in a wide range of applications, including low molecular-weight contaminant and salt removal, buffer exchange, and sample concentration.

Unlike standard flat tubing, these innovative devices do not require knots or clips that can lead to leaking and sample loss. The cassette membranes are composed of low-binding regenerated cellulose for maximum sample purity and recovery. The membrane and cassette materials are compatible with most common laboratory chemicals and buffers.

The G3 cassettes are ergonomically designed to be significantly lighter and more compact than previous generations, making them easier to handle, more practical to use, and greener for the environment. This design features a new cap for effortless sample loading and retrieval, providing visual and haptic feedback upon opening and closing to help assure sample transfer and security. Sample loading and removal is easily accomplished using a serological pipette and removal of the cap or a hypodermic needle attached to a syringe and piercing of the resealable cap. All G3 cassettes offer a built-in air chamber that provides buoyancy and vertical orientation during the dialysis process.

Slide-A-Lyzer G3 Dialysis Cassettes are available for five different sample volumes ranging from 1 to 125 mL with molecular weight cut offs (MWCOs) of 2K, 3.5K, 10K, and 20K. Slide-A-Lyzer G3 Cassettes with the 10K MWCO are also offered in electron beam-treated format for applications such as cell culture, virus purification, or sample preparation for applications requiring conditions that minimize the risk of sample contamination.

Specifications
Capacity (Metric)3 mL
MembraneCellulose Membrane, Dialysis
Product LineSlide-A-Lyzer
Purification TargetBuffer Exchange, Protein
Quantity10 cassettes
FormatCassette
MWCO10 kDa
Unit SizeEach

Frequently asked questions (FAQs)

What is the surface area of the Slide-A-Lyzer G3 Dialysis Cassettes, 10K MWCO?

The following are the surface areas for the G3 cassettes:
- 3 mL capacity (Cat. No. A52971): 12 cm^2
- 15 mL capacity (Cat. No. A52972): 28 cm^2
- 30 mL capacity (Cat. No. A52973): 50 cm^2
- 70 mL capacity (Cat. No. A52974): 100 cm^2
- 125 mL capacity (Cat. No. A52975): 110 cm^2

Find additional tips, troubleshooting help, and resources within our Protein Biology Support Centers .

Citations & References (2)

Citations & References
Abstract
A novel class of inhibitors that disrupts the stability of integrin heterodimers identified by CRISPR-tiling-instructed genetic screens.
Authors:Mattson NM,Chan AKN,Miyashita K,Mukhaleva E,Chang WH,Yang L,Ma N,Wang Y,Pokharel SP,Li M,Liu Q,Xu X,Chen R,Singh P,Zhang L,Elsayed Z,Chen B,Keen D,Pirrotte P,Rosen ST,Chen J,LaBarge MA,Shively JE,Vaidehi N,Rockne RC,Feng M,Chen CW
Journal:Nature structural & molecular biology
PubMed ID:38316881
The plasma membrane is enriched for receptors and signaling proteins that are accessible from the extracellular space for pharmacological intervention. Here we conducted a series of CRISPR screens using human cell surface proteome and integrin family libraries in multiple cancer models. Our results identified ITGAV (integrin αV) and its heterodimer ... More
Characterization and host range prediction of Staphylococcus aureus phages through receptor-binding protein analysis.
Authors:Krusche J,Beck C,Lehmann E,Gerlach D,Daiber E,Mayer C,Müller J,Onallah H,Würstle S,Wolz C,Peschel A
Journal:Cell reports
PubMed ID:40022731
Bacteriophages are crucial in bacterial communities and can be used for therapy of multidrug-resistant pathogens such as Staphylococcusaureus. However, the host range of new phages remains difficult to predict. We identified the receptor-binding proteins (RBPs) of 335 S. aureus-infecting phages, yielding 8 distinct RBP clusters. Recombinant representative RBPs of all clusters, ... More