Resina de proteína A de alta capacidad MabCaptureC™
Thermo Scientific™

Resina de proteína A de alta capacidad MabCaptureC™

Una resina de cromatografía de alto rendimiento para capturar mAbs y proteínas de fusión recombinantes etiquetadas con Fc. La resina proporciona una alta capacidad de unión específica y características de estabilidad alcalina mejoradas bajo condiciones de limpieza in situ (CIP).
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
A5303325 mL
A530315 mL
A53034100 mL
A400045642 Plates
A400045665 Columns
Número de catálogo A53033
Precio (EUR)
1.920,00
Each
Cantidad:
25 mL
Precio (EUR)
1.920,00
Each

GoPure MabCaptureC Screening Plates, 96-well, 50 μL, contain MabCaptureC High-Capacity Protein A Resin, a high-performance chromatography resin for capturing monoclonal antibodies (mAbs) and recombinant Fc-tagged fusion proteins. It provides high specific-binding capacity and enhanced alkaline-stability characteristics under clean-in-place conditions (CIP), for more robust purification and resin regeneration. These ready-to-use, 96-well spin plates offer a high-throughput format for rapid antibody purification, screening, and evaluating multiple conditions in a single plate experiment.

Features of MabCaptureC High Capacity Protein A Resin

  • High binding capacity—60 mg hIgG/mL at 5-minite residence (80 mg/mL at 10-minute residence)
  • Alkaline stability—engineered to withstand common CIP procedures with 0.2 M NaOH, enabling efficient cleaning of the chromatography resin and improving product quality
  • Animal component-free raw materials
  • Versatility—clean antibody or Fc-fusion protein preparations from cell culture supernatant, serum, or ascites
  • Stability—high-affinity Protein A is securely immobilized to high purity agarose to prevent leaching of ligand, target antibody, or base support
  • Flexible— available in multiple formats, including bulk loose resin, spin columns, and 96-well spin plates

The Protein A ligand, which is produced in yeast, is immobilized on a rigid and uniformly sized agarose bead with an average particle size of 75 μm. The uniformity of the particle size, in turn, delivers superior performance characteristics over traditional resins.

MabCaptureC High-Capacity Protein A Resin, alkaline-stable, is suitable for the purification of mAbs as well as a subpopulation of scFv's and Fab fragments that contain a VH3 domain.

Available formats

  • Resin slurries—10 mL, 50 mL, and 200 mL of 50% slurry
  • Spin columns—10 mL spin columns containing 1 mL of settled resin, supplied at 5 mL 20% slurry
  • GoPure 96-well spin plates—96-well spin plates with 20 μL or 50 μL of settled resin

Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.

Especificaciones
Capacidad de unión60 mg de HIgG/ml en tiempo de residencia de 5 min (80 mg/ml en tiempo de residencia de 10 min)
DescripciónResina de proteína A de alta capacidad, de alcalina estable
Formulación50 % de pulpa
Cantidad25 mL
Tipo de soporteAgarosa entrecruzada
Tipo de columnaAgarose Resin, Affinity
FormatoFrasco
Tipo de productoHigh Capacity Protein A Resin
Fase estacionariaProtein A
Unit SizeEach
Contenido y almacenamiento
Tras su recepción, almacenar a 4 °C

Preguntas frecuentes

Can I automate the workflow using MabCaptureC High Capacity Protein A Resin?

Yes. FPLC (Fast protein liquid chromatography) use is outlined in the instruction manual, along with compatible buffer formulations, proper flow rates, and clean-in-place protocols.

Using MabCaptureC High Capacity Protein A Resin, what downstream assays are compatible with the eluted samples?

Purified samples eluted from MabCaptureC High Capacity Protein A Resin are compatible with protein assays (Bradford, BCA, Rapid Gold, A280, etc.) and can be used for industry standard testing applications (western blotting, ELISA, immunohistochemistry, immunocytochemistry, flow cytometry, etc.).

What is the maximum operational flow velocity that can be used with MabCaptureC High Capacity Protein A Resin?

The maximum operational flow velocity with MabCaptureC High Capacity Protein A Resin is 300 cm/hr or less (150 cm/hr recommended).

What is the pH stability of MabCaptureC High Capacity Protein A Resin?

MabCaptureC High Capacity Protein A Resin is stable at pH 2-14 (short-term storage) and pH 3-14 (long-term storage).

After regeneration of MabCaptureC High Capacity Protein A Resin, is there a specific buffer in which the beads should be stored?

If the beads are not going to be re-used immediately, we recommend using 20% ethanol for long-term storage.