Product Image
Applied Biosystems™
Smart Deep™ Basecaller, 3-year license renewal
Smart Deep Basecaller is an improved basecaller for use with Sequencing Analysis Software 8. This license renews an existing licenseMás información
Have Questions?
Número de catálogoCantidad
A532161 licencia
Número de catálogo A53216
Precio (MXN)
-
Cantidad:
1 licencia
Smart Deep Basecaller is an improved basecaller for use with Sequencing Analysis Software 8. This license renews an existing license for use of Smart Deep Basecaller for 3 years.
Relative to KB Basecaller (included with Sequencing Analysis Software 8), this improved basecaller provides:
• Increased read lengths—more high quality basecalls at 5’ and 3’ ends
• More accurate pure and mixed base calls
• Improved basecalling accuracy through artifacts such as dye blobs, N-1 peaks, and mobility shifts
• Improved basecalling accuracy through heterozygous indel variants
• Reduced manual review time—fewer edits and fewer false positives
• Optional enhanced view trace visualization with a cleaned-up baseline and increased resolution in the 3’ end of plasmid (pure base) sequences
Note: SDB basecalling is enabled for data from SeqStudio Flex, SeqStudio, 3730, and 3500 series instruments.
The Smart Deep Basecaller license key is delivered by email and is then entered in Sequencing Analysis Software 8 to activate the license.
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Introducción de datosSecuenciación CE (archivos .ab1)
Licencia3 Year (Renewal)
Sistema operativoWindows 10
Tipo de productoSmart Deep Basecaller 3 Year Renewal License
Cantidad1 licencia
Software TypeSequencing Data Analysis
Unit SizeEach