Product Image
Applied Biosystems™
Variant Reporter™ Software v3.1, initial license
This is the Initial License for Variant Reporter Software. This software performs comparative sequencing, also known as direct sequencing, medicalWeitere Informationen
Have Questions?
KatalognummerMenge
A535521 Lizenz
Katalognummer A53552
Preis (EUR)
-
Menge:
1 Lizenz
This is the Initial License for Variant Reporter Software. This software performs comparative sequencing, also known as direct sequencing, medical sequencing, PCR sequencing, and resequencing with DNA sequencing files. The software is designed for reference-based and non-reference-based analysis such as mutation detection and analysis, SNP discovery and validation, and sequence confirmation. The robust algorithms will call SNPs, mutations, insertions, deletions, and heterozygous insertions/deletions for data generated using the Applied Biosystems genetic analyzers.
Nur für Forschungszwecke. Nicht zur Verwendung bei diagnostischen Verfahren.
Specifications
DateneingabeCE-Sequenzierung (.ab1-Dateien)
LizenzErstmalige Erstellung
BetriebssystemWindows 10
ProdukttypVariant Reporter Software v3.0 Initial License
Menge1 Lizenz
SoftwarekategorieUnterstützt Dateien von allen AB Genanalysatoren
ProduktlinieVariant Reporter
Software TypeAnalyse von Sequenzdaten
Unit SizeEach
Inhalt und Lagerung
The software is provided in the form of a USB stick.
Häufig gestellte Fragen (FAQ)
Where can I download the Release Notes for Variant Report Software v3.1?
Please download this PDF (http://downloads.thermofisher.com/Variant_Reporter_Software_v3.1/100107876_Variant_Reporter_v3.1_Release_Notes.pdf).
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What has been updated in Variant Reporter Software v3.1?
The primary updates for Variant Reporter Software v3.1 are the following:
- Variant Reporter Software v3.1 is supported only on Microsoft Windows 10 Professional/Enterprise operating system
- Variant Reporter Software v3.1 supports data generated from SeqStudio Flex Series Genetic Analyzers
- Variant Reporter Software v3.1 uses KB Basecaller version 1.4.2.6 for basecalling
- Variant Reporter Software v3.1 supports upgrade from Variant Reporter Software v3
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Which instrument data does Variant Reporter Software v3.1 support?
Variant Reporter Software v3.1 supports data generated from the following Applied Biosystem instruments:
- SeqStudio Flex Series Genetic Analyzers
- SeqStudio Genetic Analyzer
- 3500 Genetic Analyzer
- 3500xL Genetic Analyzer
- 3130 Genetic Analyzer
- 3130xl Genetic Analyzer
- 3730 DNA Analyzer
- 3730xl DNA Analyzer
- 310 Genetic Analyzer
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
How can I import my data from Variant Reporter Software?
For importing data, there are many ways to do so:
More information on backing up the Data Store can be found in the Variant Reporter Help Contents under the keyword “Back-up”.
1. Select the Data Store where you would like the data imported to. To choose a Data Store:
a. Click the Manage button in the Data Store section of the Dashboard View.
b. Highlight the Data Store you wish to use and click on Change Location to select it. Click Close to close the window.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
How can I archive a selected project in the Variant Reporter Software?
To archive a Variant Reporter software project from the Dashboard:
1. Open the Dashboard View.
2. In the Project tab select the project you want to archive, then click .
3. In the Archive Destination dialog box, browse to the folder where you want the project archived.
4. Click OK. The project file you archived will no longer appear in the Variant Reporter Dashboard.
More information on archiving Project data can be found in the Variant Reporter Help Contents under the keyword “Archive”.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.