Absolute Q™ Viral Titer dPCR Assays - FAQs
Absolute Q™ Viral Titer dPCR Assays - FAQs
View additional product information for Absolute Q™ Viral Titer dPCR Assays - FAQs (A53736)
4 product FAQs found
Can the Absolute Q Viral Titer assays (Cat. No. A53736) be shipped at ambient temperature?
Yes, the Absolute Q Viral Titer assays fall under our general TaqMan stability guidelines. We tested TaqMan assays at simulated summer temperatures over 4 weeks with no decrease in performance, so the assays should be safe to use if they are shipped at ambient temperature. You can refer to the following white paper for detailed testing condtions: Custom Primers and TaqMan Probes shipped at ambient temperature reduce environmental mpact and retain their quality and stability .
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.
What is the alternative for Absolute Q AAV dPCR Assay (Cat. No. A52740)?
We recommend the Absolute Q Viral Titer Assays for digital PCR (Cat. No. A53736) as an alternative to Cat. No. A52740.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.
What is the concentration of the Absolute Q Viral Titer dPCR Assays (Cat. No. A53736) and how many reactions can I get out of a single vial?
The above assays are at a 20x concentration and you can get 700 reactions from it.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.