KingFisher™ PlasmidPro Maxi Processor Endotoxin-Free Cartridge
KingFisher™ PlasmidPro Maxi Processor Endotoxin-Free Cartridge
KingFisher™ PlasmidPro Maxi Processor Endotoxin-Free Cartridge
Thermo Scientific™

KingFisher™ PlasmidPro Maxi Processor Endotoxin-Free Cartridge

The KingFisher PlasmidPro Maxi Processor Endotoxin-Free Cartridge is used with the KingFisher PlasmidPro Maxi Processor for fully automated isolation ofRead more
Have Questions?
Catalog NumberQuantity
A540724 Preps
Catalog number A54072
Price (MXN)
11,338.05
Pack of 4
Quantity:
4 Preps
Price (MXN)
11,338.05
Pack of 4

The KingFisher PlasmidPro Maxi Processor Endotoxin-Free Cartridge is used with the KingFisher PlasmidPro Maxi Processor for fully automated isolation of endotoxin-free plasmid DNA in under 75 minutes. Purify high-quality plasmid DNA that is suitable for advanced transfection applications such as transfection of sensitive cells lines or genome engineering.

Features and benefits

  • Convenient one-piece cartridge pre-filled with all necessary reagents
  • Collection tube included for simple removal and storage of plasmid DNA
  • Designed for mess-free cleanup and waste disposal
  • Plasmid DNA contains less than <0.1 EU/μg

Additional product details

The KingFisher PlasmidPro Maxi Processor is a cutting-edge solution designed for fully automated maxi-scale plasmid purification.

At the end of the procedure, the system delivers endotoxin-free plasmid DNA. The purified DNA is resuspended in TE buffer, ensuring optimal quality and compatibility with downstream applications.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Plasmid purification
For Use With (Equipment)KingFisher™ PlasmidPro Maxi Processor
Purification TargetPlasmid DNA
Quantity4 Preps
Run Time<75 min.
Sample Volume150 mL
Shipping ConditionRoom Temperature
Product TypeCartridge
Unit SizePack of 4
Contents & Storage
4 KingFisher™ PlasmidPro Maxi Processor Endotoxin-free Cartridges

Store at room temperature (15°C to 25°C).

Frequently asked questions (FAQs)

Can I resuspend a frozen pellet for use with the KingFisher PlasmidPro Maxi Processor?

When using frozen pellets as a starting point, please consider the following guidelines:

  • The pellet should represent 150 mL of the specified culture.
  • Fully resuspend the pellet in 150 mL of 1X PBS or the parent media.
  • The resuspension can be achieved by either pipetting up and down or vigorous vortexing.


Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.

What are the storage conditions for the KingFisher PlasmidPro Maxi Processor Cartridge? Is refrigeration required?

We recommend storing the KingFisher PlasmidPro Maxi Processor Cartridge at room temperature. Refrigeration is not needed.

Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.

Can I reuse the KingFisher PlasmidPro Maxi Processor Cartridge?

The KingFisher PlasmidPro Maxi Processor Cartridge is single-use and should be disposed of after use, minimizing clean-up and risk of cross-contamination. Re-use of the cartridge will contaminate and potentially harm the instrument.

Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.

Is the KingFisher PlasmidPro Maxi Processor Cartridge animal origin-free?

No. The RNaseA used in the KingFisher PlasmidPro Maxi Processor Cartridge is not animal origin-free. Review the Certificate of Analysis (COA) for more details.

Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.

What solution is the plasmid DNA sample resuspended in at the end of the purification using the KingFisher PlasmidPro Maxi Processor?

At the end of the purification using the KingFisher PlasmidPro Maxi Processor, the plasmid DNA sample is resuspended in TE buffer (10 mM Tris-HCl, pH 8.0, 0.1 mM EDTA).

Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.