The Ion Torrent Genexus Dx GX5 Chip and Genexus Coupler includes two GX5 Dx chips and two Genexus couplers, sufficientRead more
Have Questions?
Catalog Number
Quantity
A54106
2 chips
Catalog number A54106
Price (MXN)
37,560.52
Each
Quantity:
2 chips
Price (MXN)
37,560.52
Each
The Ion Torrent Genexus Dx GX5 Chip and Genexus Coupler includes two GX5 Dx chips and two Genexus couplers, sufficient for two initializations of the Genexus Dx Integrated Sequencer.
The Genexus Dx Integrated Sequencer is an in vitro diagnostic (IVD) medical device intended to be used by trained personnel in a professional laboratory environment for automated, qualitative targeted sequencing of human DNA or RNA extracted from formalin-fixed, paraffin-embedded (FFPE) tumor tissue samples or plasma samples.
For In Vitro Diagnostic Use. Not available in all countries or regions.
Specifications
For Use With (Application)Sequencing
For Use With (Equipment)Genexus™ Dx Integrated Sequencer
Product LineGenexus™ Dx
Product TypeChip and Coupler
Quantity2 chips
Shipping ConditionRoom Temperature
Software2 GX5 Dx chips
Source2 Genexus couplers
Unit SizeEach
Contents & Storage
• 2 GX5 Dx chips • 2 Genexus couplers
Store at room temperature.
Frequently asked questions (FAQs)
How could I detect if my Genexus Dx GX5 Chip and Genexus Coupler pair is faulty?
If the number of sample reads is very low, and CF-1 metrics fail QC, there could be a potential issue with the chip/coupler.
How long can Genexus Dx GX5 Chip and Genexus Coupler remain on the Genexus Dx instrument?
The chip and coupler remain installed on the instrument until all lanes are used or up to 2 weeks, whichever comes first. Once a chip and coupler set is removed from the Genexus Dx platform, it can never be reused.
How many sequencing reads are possible with Genexus Dx GX5 Chip?
The Genexus Dx GX5 Chip generates 12-15 million sequencing reads per lane, or 48-60 million reads per chip. The four-lane chip design provides maximum flexibility for sample processing, enabling sequencing of one to four lanes per run, with the in-use stability to utilize any remaining lanes within two weeks of the initial run.
What is the function of the coupler in Genexus Dx GX5 Chip and Genexus Coupler?
The coupler allows the pipettor to dispense reagents into the desired chip lane and it shuttles the chip between the loading and sequencing area on the Genexus Dx platform. Every time you change the chip, you should change the coupler.
What is contained in Genexus Dx GX5 Chip and Genexus Coupler?
The Genexus Dx GX5 Chip and Genexus Coupler (Cat. No. A54106) contains two GX5 chips and two Genexus couplers, sufficient for up to eight sequencing runs with the Genexus Dx Integrated Sequencer.