CTS™ HiFi Cas9 Protein, 5 mg - FAQs
CTS™ HiFi Cas9 Protein, 5 mg - FAQs
View additional product information for CTS™ HiFi Cas9 Protein - FAQs (A54224, A54223)
5 product FAQs found
What is CTS?
The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.
Who can I contact for outlicensing questions regarding Gibco CTS HiFi Cas9 Protein (Cat. No. A54223, A54224) and Gibco CTS TrueCut Cas9 Protein (Cat. No. A45220, A45221)?
The Limited Use Label Licenses are available on the individual product pages. For additional questions, please contact outlicensing@thermofisher.com.
Where do I find the traceability documentation for Gibco CTS HiFi Cas9 Protein (Cat. No. A54223, A54224) and Gibco CTS TrueCut Cas9 Protein (Cat. No. A45220, A45221)?
To request traceability documentation for Gibco CTS HiFi Cas9 Protein (Cat. No. A54223, A54224) and Gibco CTS TrueCut Cas9 Protein (Cat. No. A45220, A45221), please follow the instructions at the following link: https://www.thermofisher.com/us/en/home/clinical/cell-gene-therapy/cell-therapy/cell-therapy-systems/cell-therapy-systems-support.html
How is Gibco CTS HiFi Cas9 Protein (Cat. No. A54223, A54224) different from Gibco CTS TrueCut Cas9 Protein (Cat. No. A45220, A45221)?
Gibco CTS TrueCut Cas9 Protein (Cat. No. A45220, A45221) is a Streptococcus pyogenes Cas9 wild-type protein, produced by recombinant DNA technology in E. coli and Gibco CTS HiFi Cas9 Protein (Cat. No. A54223, A54224) is a Streptococcus pyogenes high-fidelity CRISPR Cas9 protein, produced by recombinant DNA technology in E. coli. Both proteins are designed and manufactured in compliance with GMP.
How is Gibco CTS HiFi Cas9 Protein (Cat. No. A54223, A54224) different from Invitrogen TrueCut HiFi Cas9 Protein (Cat. No. A50574, A50575)?
Both Gibco CTS HiFi Cas9 Protein (Cat. No. A54223, A54224) and Invitrogen TrueCut HiFi Cas9 Protein (Cat. No. A50574, A50575) are Streptococcus pyogenes high-fidelity CRISPR Cas9 proteins, produced by recombinant DNA technology in E. coli. Both proteins have the same sequence with added nuclear localization signal. Gibco CTS HiFi Cas9 Protein is manufactured to meet the standards for “Ancillary Materials for Cell, Gene, and Tissue-Based Products” whereas Invitrogen TrueCut HiFI Cas9 Protein is a research-grade product.