CTS™ AAV-MAX Transfection Kit
Gibco™

CTS™ AAV-MAX Transfection Kit

The Gibco CTS AAV-MAX Transfection Kit is an essential component of the CTS AAV-MAX Helper-Free AAV Production System.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoQuantity
A54277031 Kit (For 100 L of production)
A54277011 Kit (For 1 L of production)
A54277021 Kit (For 10 L of AAV production)
Número do catálogo A5427703
Preço (BRL)
-
Quantity:
1 Kit (For 100 L of production)

The Gibco CTS AAV-MAX Transfection Kit is an essential component of the CTS AAV-MAX Helper-Free AAV Production System. It is used for the transient transfection of CTS Viral Production Cells 2.0. The kit includes a cationic lipid-based transfection reagent, as well as a transfection booster and production enhancer, which are designed to work together to produce adeno-associated virus (AAV) particles from CTS Viral Production Cells 2.0 when cultured in CTS Viral Production Medium.

Features of the CTS AAV-MAX Transfection Kit include:

  • Components work synergistically within the CTS AAV-MAX system to produce high AAV titers greater than 5 x 1010 vg/mL (unpurified)
  • Robust transfection protocols adaptable to large scale transfection
  • Manufactured in conformity with cGMP for medical devices (21 CFR Part 820) and following USP <1043> and European Pharmacopoeia (Ph. Eur.) 5.2.12 recommendations. Drug Master File and Regulatory Support File available upon request

Components of the kit include:

  • CTS AAV-MAX Transfection Reagent — cationic lipid-based transfection reagent for helper virus-free triple transfection of high-density cultures
  • CTS AAV-MAX Booster — chemically defined component that works with Transfection Reagent to achieve high transfection efficiency
  • CTS AAV-MAX Enhancer — chemically defined component that enhances the production of AAV particles

The CTS AAV-MAX Helper-Free AAV Production System is a scalable, high-titer platform for suspension AAV production with research-grade (Cat. No. A51217) and GMP versions to enable seamless transition from research to commercial manufacturing. Other components of the CTS AAV-MAX system include CTS Viral Production Cells 2.0 (A48400), CTS Viral Production Medium (A5144001), CTS Viral-Plex Complexation Buffer (A5145401), and Viral Production Medium, AGT (A5147101).

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.
Especificações
Expression SystemMammalian
For Use With (Application)AAV Production
Phenol Red IndicatorNo
Product LineAAV-MAX
Product TypeTransfection Kit
Quantity1 Kit (For 100 L of production)
Shelf Life12 Months
FormatBag
Transfection TechniqueLipid-based Transfection, Transient Transfection
Unit SizeEach
Conteúdo e armazenamento
• 1 x 600 mL bag of CTS AAV-MAX Transfection Reagent
• 1 x 300 mL bag of CTS AAV-MAX Transfection Booster
• 1 x 1000 mL bag of CTS AAV-MAX Enhancer

Store at 2°C to 8°C. Do not freeze.