RapidINTEL™ Plus Ladder Cartridge Kit
Applied Biosystems™

RapidINTEL™ Plus Ladder Cartridge Kit

The Applied Biosystems RapidINTEL Plus Ladder Cartridge Kit is for use with the Applied Biosystems RapidHIT ID System (Cat. No. A41810).
Have Questions?
Catalog NumberQuantity
A543441 Kit (5 Cartridges)
Catalog number A54344
Price (MXN)
27,557.31
Each
Quantity:
1 Kit (5 Cartridges)
Price (MXN)
27,557.31
Each

The Applied Biosystems RapidINTEL Plus Ladder Cartridge Kit is for use with the Applied Biosystems RapidHIT ID System (Cat. No. A41810). This kit contains five RapidINTEL Plus Ladder Cartridges for use with RapidINTEL Plus Sample Cartridges.

Required for use

  • Applied Biosystems RapidHIT ID System (Cat. No. A41810)
  • RapidHIT ID Instrument Software v2.0 or higher
  • RapidHIT ID Primary Cartridge (Cat. No. A53084)

For Research Use Only.

Specifications
For Use With (Application)Human Identification
For Use With (Equipment)RapidHIT™ ID
Quantity1 Kit (5 Cartridges)
Shelf Life12 Months
FormatKit
Unit SizeEach