Thermo Scientific™
KingFisher™ SpeciTRAX™ Sample Transfer System, 1-year AB Assurance
Catalog number: A54552
Thermo Scientific™
KingFisher™ SpeciTRAX™ Sample Transfer System, 1-year AB Assurance
Catalog number: A54552
Catalog Number
A54552
Unit Size
1 package
Price (USD)
Catalog NumberUnit SizePrice (USD)
A545521 packageRequest A Quote
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific KingFisher SpeciTRAX Sample Transfer Instrument automates accessioning and sample transfer while minimizing the manual work of tube uncapping, barcode scanning, and pipetting. This innovative system combines new sample-handling automation technologies to offer ease of use, as well as increased throughput and safety, while delivering consistent, accurate results.
Features of the KingFisher SpeciTRAX instrument include:
• Transfer 192 samples from capped sample tubes to two 96-deep-well plates in 40 minutes, all within a closed-door system
• Tube decapping/recapping, barcode scanning, sample transfer
• Precision pipetting done with fluid sensing tips
• HEPA filtration and UV de-contamination
Accelerate and simplify your workflows with the KingFisher SpeciTRAX Sample Transfer System. This cutting-edge instrument combines automation technology for sample handling and robust safety features, reducing the bottleneck in sample processing and supporting increased throughput, consistency, and reproducibility of results. Integrating the KingFisher SpeciTRAX system removes tedious manual steps like decapping, recapping, and sample tracking, which can accelerate processing and save precious lab space.
For Research Use Only.
Specifications
Warranty
1 year manufacturer warranty plus 1 year service contract coverage