Applied Biosystems™ QuantStudio™ 5 Strip Plate Tray
Thermo Scientific™
Applied Biosystems™ QuantStudio™ 5 Strip Plate Tray
Setup, transfer and run the SureTect PCR assays in the QuantStudio 5 instrument using the QuantStudio 5 Strip Plate tray.
Have Questions?
Catalog NumberQuantity
A5533210
Catalog number A55332
Price (MXN)
-
Quantity:
10
Setup, transfer and run the Thermo Scientific™ SureTect™ PCR Assays in the Applied Biosystems™ QuantStudio™ 5 Food Safety Real-Time PCR System using the Applied Biosystems™ QuantStudio™ 5 Strip Plate Tray.
Designed specifically for use with the Thermo Scientific™ SureTect™ PCR Assay workflow the QuantStudio 5 Strip Plate tray is used alongside the Thermo Scientific™ 96-well PCR Base to easily setup and transfer PCR tubes into the QuantStudio 5 instrument regardless of sample throughput enabling a simple, streamlined workflow for maximum efficiency and productivity.
Use the QuantStudio 5 Strip Plate Tray to setup, transfer and run your SureTect Assays in the Quant Studio 5 instrument.
Designed to work in conjunction with the Thermo Scientific 96 Well PCR Base, the QuantStudio 5 Strip Plate Tray enables you to streamline your SureTect workflow regardless of sample throughput.
Combined with our comprehensive portfolio of assays and world-class instrumentation you can perform food safety tests confidently knowing exactly what you are getting – quality combined with simplicity.
We are committed to enabling you to experience the true confidence in your results, right-first time, every time, and the Therrmo Scientific™ SureTect™ Real-Time PCR System is continually evolving to support your laboratory and testing needs.
Specifications
DescriptionApplied Biosystems™ QuantStudio™ 5 Strip Plate Tray
For Use With (Equipment)0.1 mL block Quant Studio 5 Instrument
Quantity10
Sample TypeFood
Unit SizeEach
Contents & Storage
Room Temperature