Search
Ease of sample set up and reduced handling steps enabled using the SimpliAmp Strip Plate Tray.
| 產品號碼 | Quantity |
|---|---|
| A55333 | 10 |
Setup and transfer the SureTect lysis tubes into the SimpliAmp instrument using the Applied Biosystems™ SimpliAmp™ Strip Plate Tray.
Designed specifically for use with the Thermo Scientific™ SureTect™ PCR Assay workflow the SimpliAmp Strip Plate tray is used alongside the Thermo Scientific™ 96 Well Base to easily setup and transfer lysis tubes into the SimpliAmp instrument, regardless of sample throughput, reducing hands on steps and enabling a simple, streamlined workflow for maximum efficiency and productivity in your laboratory.
Use the SimpliAmp Strip Plate Tray to easily setup and transfer your SureTect lysis tubes into the SimpliAmp instrument.
Designed to work in conjunction with the Thermo Scientific 96 well base, the SimpliAmp Strip Plate Tray enables you to streamline your Thermo Scientific™ SureTect™ PCR Assay Workflow regardless of sample throughput.
Combined with our comprehensive portfolio of assays and world-class instrumentation you can perform food safety tests confidently knowing exactly what you are getting – quality combined with simplicity.
We are committed to enabling you to experience the true confidence in your results, right-first time, every time, and the Thermo Scientific™ SureTect™ Real-Time PCR System is continually evolving to support your laboratory and testing needs.