RapidINTEL™ Plus Sample Cartridge Evaluation Kit
Applied Biosystems™

RapidINTEL™ Plus Sample Cartridge Evaluation Kit

The Applied Biosystems RapidINTEL Plus Sample Cartridge Evaluation Kit is used with the Applied Biosystems RapidHIT ID System (Cat. No. A41810) to generate lab-quality forensic DNA profiles.
Have Questions?
Catalog NumberQuantity
A555981 Kit (10 Cartridges)
Catalog number A55598
Price (MXN)
51,646.65
Each
Quantity:
1 Kit (10 Cartridges)
Price (MXN)
51,646.65
Each

The Applied Biosystems RapidINTEL Plus Sample Cartridge Evaluation Kit is used with the Applied Biosystems RapidHIT ID System (Cat. No. A41810) to generate lab-quality forensic DNA profiles. The DNA profiles generated from RapidINTEL Plus sample cartridges can be compared to DNA profiles obtained from crime scene evidence to link one crime to another or to DNA profiles from reference samples to link a crime to an individual.

This kit contains 10 RapidINTEL Plus Sample Cartridges, each of which is an individually-wrapped, single-use, disposable cartridge that contains reagents for developing a forensic DNA profile from a single sample in 90 minutes.

The RapidINTEL 'Plus' sample cartridge is an upgraded cartridge that provides more complete, more consistent data from crime scene samples. In addition, internal quality control (IQC) markers and quantification markers have been added to assist users with identification of low quantity, degraded, and inhibited samples.

To run reference samples on the Applied Biosystems RapidHIT ID System, see our ACE GlobalFiler Sample Cartridge Kit (Cat. No. A41831 or 400114RH).

Required for use

  • Applied Biosystems RapidHIT ID System (Cat. No. A41810)
  • RapidHIT ID Instrument Software v2.0 or higher
  • RapidHIT ID Primary Cartridge (Cat. No. A53084)

For Research Use Only.

Specifications
For Use With (Application)Human Identification
For Use With (Equipment)RapidHIT™ ID
Quantity1 Kit (10 Cartridges)
Shelf Life12 Months
FormatKit
Unit SizeEach