How many samples in total can I run with Ion AmpliSeq Polyploidy Panel Kit with Ion 530 Chips (480 Samples) (Cat. No. GS45690)?
The kit is sufficient for running up to 480 samples since 8 chips are included in the kit and each chip can run up to 60 samples.<