TrueMark&trade; Amplification Control I (5 x 10<sup>7</sup> copies/&mu;L)
Applied Biosystems™

TrueMark™ Amplification Control I (5 x 107 copies/μL)

The TrueMark Amplification Control I for TrueMark plates contains a linearized multi-target plasmid pool with microbial target sequences used onRead more
Have Questions?
Catalog NumberQuantity
A5569850 μL
Catalog number A55698
Price (MXN)
46,123.15
Each
Quantity:
50 μL
Price (MXN)
46,123.15
Each
The TrueMark Amplification Control I for TrueMark plates contains a linearized multi-target plasmid pool with microbial target sequences used on TrueMark infectious disease research panels. The control has targets relevant to respiratory, urinary, vaginal, and genital microbiota research. The control can be included as a positive control for panel-specific amplification. It is available in two concentrations: 5 x 107 and 1 x 105 copies/μL. The higher concentration can be used to determine parameters necessary for analytical validation (e.g., limit of detection and Ct cut-off values).

The plasmid also contains target sequences for TaqMan Universal Extraction Control Organism (Bacillus atrophaeus) and the human RNase P gene. During real-time PCR, the control can be used as a stand-alone sample to verify assay performance and to help with troubleshooting.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatTube
Product LineTaqMan
Quantity50 μL
Shelf Life1 year
Shipping ConditionDry ice
Volume50 μL
Concentration5 x 107 Copies/μL
Detection MethodPrimer-probe
Unit SizeEach
Contents & Storage
Store at -15°C to -25°C.