Treat samples with the Thermo Scientific™ Barnstead™ FI-Streem™ Glass Stills Pretreatment Kit when feed water quality does not match system requirements.
Have Questions?
Change view
Catalog Number
Description
A56285
Pretreatment Kit, 120V
A56280
Pretreatment Kit, 240V
2 Options
Catalog number A56285
Price (MXN)
-
Description:
Pretreatment Kit, 120V
Treat samples with the Thermo Scientific™ Barnstead™ FI-Streem™ Glass Stills Pretreatment Kit when feed water quality does not match system requirements.
Utilizes disposable cartridges
D × W × H: 8.28 × 11 × 23.5in. (21 × 27.9 × 59.7cm)
Specifications
DescriptionPretreatment Kit, 120V
For Use With (Equipment)Barnstead FI-Streem Glass Still