SureTect™ Salmonella Infantis PCR Assay
Thermo Scientific™

SureTect™ Salmonella Infantis PCR Assay

Rapidly detect Salmonella Infantis in carcass rinses and poultry part samples with the Thermo Scientific™ SureTect™ Salmonella Infantis PCR Assay workflow.

Have Questions?
Catalog NumberNo. of Tests
A5645096 tests/kit
Catalog number A56450
Price (USD)
-
No. of Tests:
96 tests/kit

Rapidly detect Salmonella Infantis in carcass rinses and in up to 375 g poultry parts samples with the Thermo Scientific™ SureTect™ Salmonella Infantis PCR Assay, which incorporates optimized primers and probe components for highly specific results.

Designed specifically for Salmonella Infantis control programs in poultry production, the SureTect Salmonella Infantis PCR Assay has been extensively tested and verified on poultry matrices from food manufacturing environments and addresses process challenges by offering accuracy and efficiency in product release or intervention.

Combine Thermo Scientific™ SureTect™ Salmonella species, Typhimurium and Enteritidis Multiplex PCR Assay with the SureTect Salmonella Infantis PCR Assay to simultaneously detect Salmonella spp. and identify the top 3 serotypes in poultry products linked to human illness: S. Typhimurium, S. Enteritidis, and S. Infantis.

The SureTect PCR System is an optimized solution combining speed and performance in an easy-to-use, cost-effective platform—giving you results you can trust.

Simple

  • Easy-to-use and simple workflow
  • Fast screening and identification of positive samples
  • All-in-one workflow to get same-time results from a single test portion: Salmonella spp. and top 3 serotypes (S. Enteritidis, S. Typhimurium, and S. Infantis)
  • Pre-dispensed and lyophilized reagents

Fast and Flexible

  • Option to automate the lysis and PCR sample preparation
  • Optimized workflow delivers on-screen results in as little as 18 hours
  • Flexibility to run SureTect Salmonella species, Typhimurium and Enteritidis Multiplex PCR Assay with SureTect Salmonella Infantis Assay, simultaneously
  • Test up to 96 samples in each run
  • Choice of sealing lysis tubes with pierceable seals or caps
  • Automatic lysis cycle
  • Open PCR platforms

Smart

  • Intuitive, user-friendly software delivers reliable results, with no subjective interpretation
  • Actionable data supports fast decisions in a complex product supply chain
  • IPC in every assay for complete confidence in results

The Thermo Scientific™ SureTect™ Salmonella Infantis PCR Assay Kit includes:

  • Lysis Reagent 1 Tubes
  • Proteinase K
  • Salmonella Infantis PCR Tubes
  • Thermo Scientific 96-Well Pierceable Seals
  • PCR Caps

The SureTect Salmonella Infantis PCR Assay is validated on the Applied Biosystems™ QuantStudio™ 5 Food Safety Real-Time PCR System with RapidFinder™ Analysis Software v3.0 or later or the Applied Biosystems™ 7500 Fast Food Safety 96-well Real-Time PCR instrument using Rapidfinder™ Express Software v2.0 or later, allowing you to test easily and confidently. After completing the PCR run, interpreted results are clearly displayed and can be reported, stored, printed, or downloaded as required.

For Testing of Food and Environmental Samples Only.
Specifications
Certification ComplianceISO 9001
Control TemplateInternal Positive Control (IPC)
DiseaseSalmonellosis
Final Product TypeSalmonella Infantis DNA
For Use With (Equipment)Real-Time PCR (qPCR) System
FormatKit
High-throughput CompatibilityHigh-throughput Compatible, Automated Protocols
Hot StartBuilt-In Hot Start
IncludesLysis Reagent 1 Tubes (12 strips of 8 tubes), Proteinase K (1 tube), Salmonella Infantis PCR Tubes (12 strips of 8 tubes, 1 pellet each), Thermo Scientific 96-Well Pierceable Seals (2 sheets), PCR Caps (12 strips of 8 caps)
Includes Label or DyeYes
Kit TypeqPCR Salmonella Infantis Detection Kit
No. of Tests96 tests/kit
PrimerPCR Primers
Product LineSureTect
Product TypeSureTect Salmonella Infantis PCR
Reaction FormatSeparate Components
Reagent TypePCR Assay
Research CategoryFood Safety
Sample TypeRaw Poultry Parts, Poultry Carcass Rinse
Starting MaterialFood Samples or Production Environment Samples
Sufficient For96 Tests
Target Organism ClassSalmonella Infantis
TechniqueReal-time PCR
Volume10 μL
Detection MethodqPCR, RT-PCR
For Use With (Application)Pathogen Detection
FormLyophilized
PCR MethodqPCR
Unit SizeEach
Contents & Storage
Store at 2 to 8°C