iBlot™ 3 Western Blot Transfer Device
iBlot™ 3 Western Blot Transfer Device
iBlot™ 3 Western Blot Transfer Device
iBlot™ 3 Western Blot Transfer Device
Invitrogen™

iBlot™ 3 Western Blot Transfer Device

The iBlot 3 Western Blot Transfer Device is a dry transfer device that performs western blotting transfer efficiently and reliably in as few as three minutes and without the need for liquid buffers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWarranty
A56727REX Extended Warranty, 2 years
IB31001Standard, 1-year warranty included
Catalog number A56727
Price (USD)
4,889.39
Each
Warranty:
REX Extended Warranty, 2 years
Price (USD)
4,889.39
Each

The iBlot 3 Western Blot Transfer Device is a dry transfer device that performs western blotting transfer efficiently and reliably in as few as three minutes and without the need for liquid buffers. The iBlot 3 system is compatible with both nitrocellulose and polyvinylidene difluoride (PVDF) membranes and offers equivalent or better performance compared with traditional wet-transfer methods in a fraction of the time. The iBlot 3 Western Blot Transfer Device is an integral part of the iBlot dry blotting system, which consists of the transfer device and consumable transfer stacks that contain the required buffers and transfer membrane (nitrocellulose or PVDF).

iBlot 3 Western Blot Transfer Device features include:

  • Complete protein transfer in as few as three minutes
  • High detection sensitivity and even, consistent protein transfer
  • Two independently controlled transfer stations capable of transferring up to two midi or four mini blots
  • Built-in adjustable cooling to ensure consistent transfer temperature
  • Pre-programed transfer methods optimized for low, high, and broad molecular weight ranges
  • Ability to create custom programs
  • A simple, user-friendly interface

How the iBlot dry blotting system works
Buffer ion reservoirs are incorporated into the gel matrix of transfer stacks instead of buffer tanks or soaked filter papers. The high density of ions in the gel matrix enables rapid protein transfer. During blotting, the copper anode does not generate oxygen gas as a result of water electrolysis, reducing blot distortion (conventional protein transfer techniques, including wet, semi-wet, and semi-dry, use inert electrodes that generate oxygen).

With the iBlot dry blotting system, transfer time is reduced by the shortened distance between electrodes, high field strength, and high current. Trapped air bubbles, often created during the manual preparation of the blotting sandwich layers, are easy to avoid due to our unique de-bubbling design that promotes even and complete transfer. With the iBlot 3 system, there is no need to prepare buffers or clean up after blotting. The total preparation and run time is normally less than ten minutes per blot.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Membrane CompatibilityNitrocellulose, PVDF
Mode of TransferDry
Quantity1 device with warranty
Throughput2 Midi or 4 Mini Gels
WarrantyREX Extended Warranty, 2 years
For Use With (Application)Western Blot
For Use With (Equipment)iBlot 3 Transfer Stacks
Gel CompatibilityMini- or Midi-sized: Bis-Tris Gels, Tricine Gels, Tris-Acetate Gels, and Tris-Glycine Gels
Product LineiBlot 3
Running Time∼3 to 10 min.
TypeTransfer Device
Unit SizeEach
Package Components
iBlot™ 3 Western Blot Transfer Device
Catalog Number
IB31001
Unit Size
1 device
Qty
1
ZGEXSCIBLOT3
Catalog Number
ZGEXSCIBLOT3
Unit Size
Qty
1
Contents & Storage
• iBlot 3 Gel Transfer Device
• Western Blot Roller
• REX Extended Warranty

Store at room temperature.

Frequently asked questions (FAQs)

Are the E-PAGE Gels (example Cat. No. EP09606) compatible with the iBlot3 device (Cat. O. IB31001)?

Due to the thickness of the E-PAGE gels, they are not compatible with the iBlot3 device for transfer. Recommended methods for transfer of E-PAGE gels are semi-dry blotting or semi-wet blotting in the XCell II Blot Module. Methods are described in the E-PAGE Technical Guide.

Find additional tips, troubleshooting help, and resources within our Protein Gel Electrophoresis Chambers, Power Supplies, and Accessories Support Center.

Do you offer additional years of service for the iBlot 3 Western Transfer Device?

No, we do not provide service contracts or extended warranties for these devices. The warranty is limited to 1 year from the date of shipment.