iBlot™ 3 Starter Kit, NC
iBlot™ 3 Starter Kit, NC
iBlot™ 3 Starter Kit, NC
iBlot™ 3 Starter Kit, NC
Invitrogen™

iBlot™ 3 Starter Kit, NC

The iBlot 3 Starter Kit includes the iBlot 3 Western Blot Transfer Device and two boxes of iBlot 3 Transfer Stacks, Midi, NC (nitrocellulose)—everything needed to perform a rapid dry-transfer of proteins from a gel to a membrane.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWarranty
A56728REX Extended Warranty, 2 years
IB31001SStandard, 1-year warranty
Catalog number A56728
Price (USD)
5,129.42
Each
Warranty:
REX Extended Warranty, 2 years
Price (USD)
5,129.42
Each

The iBlot 3 Starter Kit includes the iBlot 3 Western Blot Transfer Device and two boxes of iBlot 3 Transfer Stacks, Midi, NC (nitrocellulose)—everything needed to perform a rapid dry-transfer of proteins from a gel to a membrane.

iBlot 3 Western Blot Transfer Device
The iBlot 3 Western Blot Transfer Device is a dry transfer device that performs western blotting transfer efficiently and reliably in as few as three minutes and without the need for liquid buffers. The iBlot 3 system is compatible with both nitrocellulose and polyvinylidene difluoride (PVDF) membranes and offers equivalent or better performance compared with traditional wet-transfer methods in a fraction of the time. The iBlot 3 Western Blot Transfer Device is an integral part of the iBlot dry blotting system, which consists of the transfer device and consumable transfer stacks that contain the required buffers and transfer membrane (nitrocellulose or PVDF).

iBlot 3 Western Blot Transfer Device features include:

  • Complete protein transfer in as few as three minutes
  • High detection sensitivity and even, consistent protein transfer
  • Two independently controlled transfer stations capable of transferring up to two midi or four mini blots
  • Built-in adjustable cooling to ensure consistent transfer temperature
  • Pre-programed transfer methods optimized for low, high, and broad molecular weight ranges
  • Ability to create custom programs
  • A simple, user-friendly interface

How the iBlot dry blotting system works
Buffer ion reservoirs are incorporated into the gel matrix of transfer stacks instead of buffer tanks or soaked filter papers. The high density of ions in the gel matrix enables rapid protein transfer. During blotting, the copper anode does not generate oxygen gas as a result of water electrolysis, reducing blot distortion (conventional protein transfer techniques, including wet, semi-wet, and semi-dry, use inert electrodes that generate oxygen).

With the iBlot dry blotting system, transfer time is reduced by the shortened distance between electrodes, high field strength, and high current. Trapped air bubbles, often created during the manual preparation of the blotting sandwich layers, are easy to avoid due to our unique de-bubbling design that promotes even and complete transfer. With the iBlot 3 system, there is no need to prepare buffers or clean up after blotting. The total preparation and run time is normally less than ten minutes per blot.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Membrane CompatibilityNitrocellulose, PVDF
Mode of TransferDry
Quantity1 kit with warranty
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
Throughput2 Midi or 4 Mini Gels
WarrantyREX Extended Warranty, 2 years
For Use With (Application)Western Blot
For Use With (Equipment)iBlot 3 Transfer Stacks
Gel CompatibilityMini- or Midi-sized: Bis-Tris Gels, Tricine Gels, Tris-Acetate Gels, and Tris-Glycine Gels
Product LineiBlot 3
Running Time∼3 to 10 min.
TypeStarter Kit
Unit SizeEach
Package Components
iBlot™ 3 Starter Kit, nitrocellulose
Catalog Number
IB31001S
Unit Size
1 kit
Qty
1
ZGEXSCIBLOT3
Catalog Number
ZGEXSCIBLOT3
Unit Size
Qty
1
Contents & Storage
• iBlot 3 Gel Transfer Device
• Western Blot Roller
• iBlot 3 Transfer Stacks, midi, nitrocellulose (2 boxes, 20 stacks)
• REX Extended Warranty

Store at room temperature.

Frequently asked questions (FAQs)

Do you offer additional years of service for the iBlot 3 Western Transfer Device?

No, we do not provide service contracts or extended warranties for these devices. The warranty is limited to 1 year from the date of shipment.