Search
| Catalog Number | Target Organism Class |
|---|---|
| A56835 | Campylobacter jejuni, C. coli and C. lari |
Rapidly detect Campylobacter jejuni, C. coli and C. lari in poultry matrices and relevant environmental samples in as little as 24 hours with the AOAC Performance Tested MethodsSM certified Thermo Scientific™ SureTect™ Campylobacter jejuni, C. coli and C. lari PCR Assay.
The improved SureTect assay format incorporates color-coded plastics with orientation markings and breakable strip format, colored reagents, and pierceable seals to enable an optimized workflow for maximum laboratory efficiency and productivity.
The SureTect System is an optimized solution combining speed and performance in an easy-to-use, cost-effective platform—giving you results you can trust.
Simple
Fast and Flexible
Smart
The Thermo Scientific™ SureTect™ Campylobacter jejuni, C. coli and C. lari PCR Assay includes:
Enriched samples are pipetted into pre-filled lysis tubes, along with Proteinase K, before incubation, to lyse any bacterial cells present in the sample and release their DNA into the solution. The lysate is then loaded into the SureTect Campylobacter jejuni, C. coli and C. lari PCR tube to re-hydrate the pre-loaded PCR pellet which contains all the necessary components and reagents for PCR, including a probe, primers, and DNA template for the internal positive control (IPC).
The PCR tubes are then sealed, loaded into the Applied Biosystems™ QuantStudio™ 5 Food Safety Real Time PCR instrument with RapidFinder™ Analysis Software v3.0 (or later) or the Applied Biosystems 7500 Fast Food Safety 96-well Real-Time PCR instrument with RapidFinder™ Express Software v2.0 (or later).
After completing the run, interpreted results are clearly displayed and can be reported, stored, printed, or downloaded as required. The internal positive control confirms correct assay conditions have occurred.
Not all products are available for sale in all territories. Please inquire.