Search
| 카탈로그 번호 | 수량 |
|---|---|
| A56848 | 96 reactions |
Rapidly and simultaneously detect, quantify, and differentiate Salmonella species, Salmonella Typhimurium, and Salmonella Enteritidis in a single real-time PCR test with the Thermo Scientific™ SureCount™ Salmonella species, Typhimurium and Enteritidis Multiplex PCR Kit. Replace labor and time-consuming traditional enumeration methods with SureCount™ rapid workflows for meat, poultry, and pork matrices. Understand Salmonella intervention effectiveness and make risk-based food safety decisions with fully automated Salmonella quantification and serotype differentiation results in as little as 8 hours using the Thermo Scientific™ RapidFinder™ Analysis Software version 3.0 or later. The kit is compatible with the Applied Biosystems™ Quant Studio™ 5 Food Safety PCR System.
Increase workflow efficiency and gain greater insight into Salmonella control across the production chain by replacing traditional enumeration and strain typing methods with the SureCount™ Salmonella Species, Typhimurium and Enteritidis Multiplex PCR method. The real-time PCR kit enables rapid, simultaneous detection and quantitation of Salmonella species, Salmonella Typhimurium, and Salmonella Enteritidis in a single PCR reaction, with workflows for a wide variety of matices including poultry, beef, and pork matrices including poultry and meat finished goods workflows (AOAC Performance Tested MethodsSM certified), and custom protocols available for in-process carcass rinses and primary production samples.
SureCount™ Salmonella Species, Typhimurium and Enteritidis Multiplex PCR Kit includes: