Thermo Scientific™ Lysis Caps, Domed
Thermo Scientific™
Thermo Scientific™ Lysis Caps, Domed
Compatible with the lysis tubes in the SureTect Real Time PCR system for pathogen detection, the domed lysis caps enable maximum flexibility for sample processing in your laboratory.
Have Questions?
Número de catálogo A56895
Precio (MXN)
-
Thermo Scientific™ Lysis Lysis Caps, Domed, strips of 8, are compatible with the 96-well SureTect Lysis Plates as an alternative to Thermo Scientific™ 96-well Piercable Seals. The cap strips have large end tabs for easy strip removal and are compatible for use with the Thermo Scientific™ Capping and Uncapping Tools.
Combined with our comprehensive portfolio of assays and world-class instrumentation, you can perform food safety tests confidently knowing exactly what you are getting – quality combined with simplicity.
We are committed to enabling you to experience the true confidence in your results, right-first time, every time, and the Thermo Scientific™ SureTect™ Real-Time PCR System is continually evolving to support your laboratory and testing needs.
Únicamente para las pruebas de muestras alimentarias y ambientales.
Especificaciones
DescripciónLysis Caps, domed
Para utilizar con (aplicación)Food & Beverage
Tipos de placasCompatible with the 96-well SureTect Lysis Plates
Cantidad120
Unit SizeEach
Contenido y almacenamiento
Room Temperature