Is the Pierce Dilution-Free BSA Protein Standards, Multichannel Pipette Compatible available as a standalone item?
Yes, we offer the Pierce Dilution-Free BSA Protein Standards, Multichannel Pipette Compatible in two versions:
- Cat. No. A56979 (0.125-10 mg/mL) for the Pierce Dilution-Free Rapid Gold BCA Protein Assay Kit (Cat. No. A55860, A55861, A55862)
- Cat. No. A55863 (0.125-2 mg/mL) which is compatible with the Pierce BCA Protein Assay Kit (Cat. No. A55864, A55865) and with most protein assays that have a working range between 0-2 mg/mL
Find additional tips, troubleshooting help, and resources within our Protein Assays and Quantitation Support Center.
- Cat. No. A56979 (0.125-10 mg/mL) for the Pierce Dilution-Free Rapid Gold BCA Protein Assay Kit (Cat. No. A55860, A55861, A55862)
- Cat. No. A55863 (0.125-2 mg/mL) which is compatible with the Pierce BCA Protein Assay Kit (Cat. No. A55864, A55865) and with most protein assays that have a working range between 0-2 mg/mL
Find additional tips, troubleshooting help, and resources within our Protein Assays and Quantitation Support Center.