CTS™ Detachable Dynabeads™ CD8 - FAQs
CTS™ Detachable Dynabeads™ CD8 - FAQs
View additional product information for CTS™ Detachable Dynabeads™ CD8 - FAQs (A56995)
16 product FAQs found
What is the definition of superparamagnetic, and what does this mean for my cell isolation application with Dynabeads magnetic beads?
Superparamagnetic means that the Dynabeads magnetic beads exhibit magnetic properties when placed within a magnetic field, but have no residual magnetism when removed from the magnetic field.
This means that your targeted cells, proteins, or nucleic acids are only subjected to magnetic forces during the time the beads are on the magnet. The beads do not aggregate, but remain evenly dispersed in suspension.
Find additional tips, troubleshooting help, and resources within our Dynabeads Cell Isolation and Expansion Support Center.
What is CTS?
The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.
What are the shipping and storage and conditions for CTS Detachable Dynabeads CD8 (Cat. No. A56995)?
CTS Detachable DynabeadsCD3/CD28 (Cat. No. A56996) is shipped at 2-8 degrees C. We recommend storing it at 2-8 degrees C, protected from light.
Find additional tips, troubleshooting help, and resources within our Dynabeads Cell Isolation and Expansion Support Center.
What are the antibodies on the surface of CTS Detachable Dynabeads CD4 and CTS Detachable Dynabeads CD8?
CTS Detachable Dynabeads CD4 and CTS Detachable Dynabeads CD8 are coated with camelid-derived, single-domain (VHH) antibody fragments that identify and bind CD4 or CD8 markers, respectively. These antibodies are produced in an Animal Origin Free (AOF) yeast-based system virtually eliminating any risk of viral contamination.
Find additional tips, troubleshooting help, and resources within our Dynabeads Cell Isolation and Expansion Support Center.
When using CTS Detachable Dynabeads CD4 or CTS Detachable Dynabeads CD8, what is the recommended ratio of Dynabeads to target T cells?
When using CTS Detachable Dynabeads CD4 or CTS Detachable Dynabeads CD8, we recommend a 4:1 ratio of Dynabeads to target T cells (CD4+ or CD8+ T cells, respectively). Please do not calculate it based on the number of total nucleated cells in your starting material.
Find additional tips, troubleshooting help, and resources within our Dynabeads Cell Isolation and Expansion Support Center.
What is the shelf life for CTS Detachable Dynabeads CD4 and CTS Detachable Dynabeads CD8?
At the time of launch to market for CTS Detachable Dynabeads CD4 and CTS Detachable Dynabeads CD8 in October 2024, the shelf life was 12 months, but our plan is to extend the shelf life to 18, 24, 36, 48, and 60 months on a rolling basis. Therefore, our expectation is that the shelf life will be 5 years by the year 2029
Find additional tips, troubleshooting help, and resources within our Dynabeads Cell Isolation and Expansion Support Center.
.
Can I use CTS Detachable Dynabeads CD4 and CTS Detachable Dynabeads CD8 together to isolate both CD8 positive and CD8 positive T cells?
Yes, CTS Detachable Dynabeads CD4 and CTS Detachable Dynabeads CD8 can be added at the same time to allow isolation of both CD4+ and CD8+ T cells.
Find additional tips, troubleshooting help, and resources within our Dynabeads Cell Isolation and Expansion Support Center.
Are CTS Dynabeads and CTS Detachable Dynabeads for clinical trial and commercial manufacturing use?
Yes, CTS Dynabeads and CTS Detachable Dynabeads are GMP-grade and CTS-branded, which means that they are eligible for use in process development, clinical trials, and commercial manufacturing.
Thermo Fisher Scientific’s Cell Therapy Systems (CTS) translates to cGMP manufacturing, testing, and traceability documentation (such as Drug Master Files and Regulatory Support Files) for customer filing needs, and a proven track record of use in many clinical trials and commercialized drugs on the market.
The CTS Dynabeads portfolio, specifically, has been actively used in over 200 clinical trials in 2023 as well as in a number of FDA-approved drugs for commercial use.
Find additional tips, troubleshooting help, and resources within our Dynabeads Cell Isolation and Expansion Support Center.
How does the active release mechanism work for CTS Detachable Dynabeads?
CTS Detachable Dynabeads are meant to be used with the CTS Detachable Dynabeads Release Buffer that comes in a bioprocess container compatible with our CTS DynaCellect Magnetic Separation System for automated workflows.
After the isolation step, the Dynabeads-bound T cells are incubated with the CTS Detachable Dynabeads Release Buffer to actively detach the cells from the Dynabeads. The CTS Detachable Dynabeads Release Buffer contains d-biotin that outcompetes a biotin derivative on the surface of CTS Detachable Dynabeads. It is not an enzymatic reaction.
Users may then rely on the CTS DynaCellect Magnetic Separation System to magnetically separate CTS Detachable Dynabeads from target T cells so they are left with a highly pure target T cell population in the positive fraction.
Find additional tips, troubleshooting help, and resources within our Dynabeads Cell Isolation and Expansion Support Center.
Are CTS Detachable Dynabeads compatible with any cell culture media?
Due to the presence of biotin in the CTS Detachable Dynabeads Release Buffer, we do not recommend using CTS Detachable Dynabeads with media that already contain high amounts of biotin, such as RPMI media. Instead, we recommend pairing with media such as our CTS OpTmizer SFM and CTS OpTmizer Pro SFM.
Note: Internal studies have shown that the biotin in the CTS Detachable Dynabeads Release Buffer does not have any impact on cell phenotype or viability.
Find additional tips, troubleshooting help, and resources within our Dynabeads Cell Isolation and Expansion Support Center.
Are Dynabeads compatible with centrifugation?
Dynabeads are not designed or intended to be used with centrifugation. Pressure from centrifugal forces should not theoretically be a problem for the Dynabeads themselves due to their rigid nature and composition. However, the force exerted by the Dynabeads on surrounding cells within the pellet may be detrimental to cell viability.
Find additional tips, troubleshooting help, and resources within our Dynabeads Cell Isolation and Expansion Support Center.
What makes Dynabeads magnetic?
Dynabeads magnetic beads are uniform, non-porous, superparamagnetic, monodispersed and highly crosslinked polystyrene microspheres consisting of an even dispersion of magnetic material throughout the bead. The magnetic material within the Dynabeads magnetic beads consists of a mixture of maghemite (gamma-Fe2O3) and magnetite (Fe3O4). The iron content (Fe) of the beads is 20% by weight in our M-450 Dynabeads.
Dynabeads magnetic beads are also coated with a thin polystyrene shell which encases the magnetic material and prevents any leakage from the beads or trapping of ligands in the bead interior. The shell also protects the target from exposure to iron while providing a defined surface area for the adsorption or coupling of various molecules.
The uniformity of Dynabeads' size and shape provides consistent physical and chemical properties. These uniform physical characteristics lead to high-quality, reproducible results.
Find additional tips, troubleshooting help, and resources within our Dynabeads Cell Isolation and Expansion Support Center.
When using CTS Detachable Dynabeads, what is the level of residual Dynabeads left after the bead removal step?
This depends on the total number of cells that you are processing in a single run and the total number of Dynabeads used. However, our large-scale studies with 10-liter volumes on the CTS DynaCellect Magnetic Separation System yielded less than 25 residual Dynabeads per 3 million cells. And with 1-liter volumes, performance was even better as we observed less than 5 residual Dynabeads per 3 million cells (again in internal studies).
As a reference point, most regulatory agencies have an unofficial minimum threshold of 100 residual beads per 3 million cells.
Find additional tips, troubleshooting help, and resources within our Dynabeads Cell Isolation and Expansion Support Center.
Can I try out CTS Detachable Dynabeads but do so manually with columns, and not with the CTS DynaCellect Magnetic Separation System?
All of our CTS Detachable Dynabeads products are intended to be used with the CTS DynaCellect Magnetic Separation System. While small DynaMag magnets can be used to assess the functionality of the product in a laboratory scale, it does not allow you to see the full benefits or accurate performance should you decide to scale up to clinical or commercial manufacturing.
Using our CTS Detachable Dynabeads without the CTS DynaCellect Magnetic Separation System is not representative of the yield and purity obtained in the automated closed system.
Find additional tips, troubleshooting help, and resources within our Dynabeads Cell Isolation and Expansion Support Center.
What is the level of support offered when using CTS Dynabeads and CTS Detachable Dynabeads in clinical trials and commercial manufacturing?
Our CTS Dynabeads and CTS Detachable Dynabeads are supported with Drug Master Files (DMFs), Regulatory Support Files (RSFs), and other traceability documentation.
We have dedicated resources from our Quality and Regulatory teams to support customers who enter into a Commercial Supply Agreement with us.
Find additional tips, troubleshooting help, and resources within our Dynabeads Cell Isolation and Expansion Support Center.