Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          1. Home▸
          2. Shop All Products▸
          3. PCR Equipment and Supplies▸
          4. PCR Supplies▸
          5. Real Time PCR Products▸
          6. Real Time PCR Reagents and Kits▸
          7. TrueMark™ STI Amplification Control
          Request Quote
          Share
          TrueMark™ STI Amplification Control
          Applied Biosystems™

          TrueMark™ STI Amplification Control

          The TrueMark STI Amplification Control contains a linearized multi-target plasmid with the following targets: Chlamydia trachomatis, Neisseria gonorrhoeae, Mycoplasma genitalium,Read more
          Have Questions?
          Catalog NumberQuantity
          A5700925 μL
          Catalog number A57009
          Price (USD)
          1,028.00
          Each
          Quantity:
          25 μL
          Price (USD)
          1,028.00
          Each
          The TrueMark STI Amplification Control contains a linearized multi-target plasmid with the following targets: Chlamydia trachomatis, Neisseria gonorrhoeae, Mycoplasma genitalium, Trichomonas vaginalis, and the human RNaseP gene.

          The amplification control has a concentration of 1 x 105 copies/μL. It can be used as a positive control for STI panel-specific amplification or as a stand-alone sample to verify assay performance and to help with troubleshooting.

          Specifications
          Quantity25 μL
          Detection MethodPrimer-probe
          For Use With (Application)Pathogen Detection
          Unit SizeEach

          Frequently asked questions (FAQs)

          Is there any cross-reactivity reported for the TrueMark STI Select Panel, Combo Kit?

          Cross-reactivity was observed with the following organisms when using the TrueMark STI Select Panel, Combo Kit:
          Organism Pentatrichomonas hominis -> Cross-reactive target: Trichomonas vaginalis
          Organism Trichomonas tenax -> Cross-reactive target: Trichomonas vaginalis

          What are the reporter dyes, targets, and quenchers used in the TrueMark STI Select Panel, Combo Kit?

          Please refer to Table 1 on page 4 of the user guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0028493-TrueMarkSTISelectPanel-UG-RUO-EN.pdf) for dyes, targets, and quenchers used in the TrueMark STI Select Panel, Combo Kit.

          What specimen types can be used with the TrueMark STI Select Panel, Combo Kit?

          Vaginal and genital swabs can be used with the TrueMark STI Select Panel, Combo Kit.

          What is the approximate workflow time for running the TrueMark STI Select Panel, Combo Kit assay starting from sample preparation?

          It takes approximately 3 hrs for running the TrueMark STI Select Panel, Combo Kit assay, starting from sample preparation to obtaining the Ct value.

          With the TrueMark STI Select Panel, Combo Kit and TrueMark STI Amplification Control, how long can I hold the reagents/reaction before starting the procedure?

          TrueMark STI Select Panel: Once thawed, the assay is stable for up to 8 hrs at 2-8 degrees C. Do not exceed 5 freeze-thaws.
          TrueMark STI Amplification Control: Once thawed, the amplification control should be used immediately and returned to -30 to -10 degrees C. Do not exceed 5 freeze-thaws.
          Assembled RT‑PCR reaction plate: The assembled RT-PCR reaction plate is stable for up to 60 mins at 2-8 degrees C.

          View all TrueMark™ STI Amplification Control FAQs
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States